Skip to Content
Merck
All Photos(1)

Key Documents

890895C

Avanti

18:1 TAP (DOTAP, MS Salt)

Avanti Research - A Croda Brand 890895C

Synonym(s):

1,2-dioleoyl-3-trimethylammonium-propane (methyl sulfate salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$368.00
200 MG
$760.00

$368.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$368.00
200 MG
$760.00

About This Item

Empirical Formula (Hill Notation):
C43H83NO8S
CAS Number:
Molecular Weight:
774.19
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$368.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (890895C-25mg)
pkg of 2 × 4 mL (890895C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890895C

concentration

10 mg/mL (890895C-25mg)
25 mg/mL (890895C-200mg)

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.O=S([O-])(OC)=O

InChI

1S/C42H80NO4.CH4O4S/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-41(44)46-39-40(38-43(3,4)5)47-42(45)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2;1-5-6(2,3)4/h20-23,40H,6-19,24-39H2,1-5H3;1H3,(H,2,3,4)/q+1;/p-1/b22-20-,23-21-;

InChI key

RSMRWWHFJMENJH-LQDDAWAPSA-M

Biochem/physiol Actions

DOTAP (1,2-dioleoyl-3-trimethylammonium-propane) is a cationic lipid, used for in vitro and in vivo nucleic acid and protein delivery.[1] The lipid composition and cell type affects the transfection potential of DOTAP.[2] DOTAP mediates gene knockdown technique in hematopoietic stem cells (HSCs).[3]

Packaging

5 mL Clear Glass Sealed Ampule (890895C-200mg)
5 mL Clear Glass Sealed Ampule (890895C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient siRNA delivery by the cationic liposome DOTAP in human hematopoietic stem cells differentiating into dendritic cells
Martino S, et al.
BioMed Research International, 2009 (2009)
DOTAP (and other cationic lipids): chemistry, biophysics, and transfection
Simberg D, et al.
Critical Reviews in Therapeutic Drug Carrier Systems, 21(4) (2004)
DOTAP/DOPE ratio and cell type determine transfection efficiency with DOTAP-liposomes
Kim BK, et al.
Biochimica et Biophysica Acta - Biomembranes, 1848(10), 1996-2001 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service