Skip to Content
Merck
All Photos(1)

Key Documents

890850C

Avanti

18:1 DAP

Avanti Research - A Croda Brand 890850C

Synonym(s):

1,2-dioleoyl-3-dimethylammonium-propane (DODAP)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$368.00
200 MG
$760.00

$368.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 MG
$368.00
200 MG
$760.00

About This Item

Empirical Formula (Hill Notation):
C41H77NO4
CAS Number:
Molecular Weight:
648.05
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$368.00


In StockDetails


Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (890850C-25mg)
pkg of 2 × 4 mL (890850C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890850C

concentration

10 mg/mL (890850C-25mg)
25 mg/mL (890850C-200mg)

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(CN(C)C)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C41H77NO4/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-40(43)45-38-39(37-42(3)4)46-41(44)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2/h19-22,39H,5-18,23-38H2,1-4H3/b21-19-,22-20-

InChI key

NYDLOCKCVISJKK-WRBBJXAJSA-N

General description

18:1 DAP is a cationic lipid. It is a pH sensitive transfection reagent, used for the preparation of cationic liposomes.[1]

Application

18:1 DAP is suitable :
  • as a component of lipid bilayers[2]
  • in the formulation of stable nucleic acid lipid vesicles[3]
  • in the preparation and physicochemical characterization of antisense oligodeoxyribonucleotide (ODN)-containing folate (FA)-targeted or non-targeted liposomes[1]

Packaging

5 mL Clear Glass Sealed Ampule (890850C-200mg)
5 mL Clear Glass Sealed Ampule (890850C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transferrin-conjugated SNALPs encapsulating 2?-O-methylated miR-34a for the treatment of multiple myeloma
Scognamiglio I, et al.
BioMed Research International, 2014 (2014)
Preparation and in-vitro evaluation of an antisense-containing cationic liposome against non-small cell lung cancer: a comparative preparation study
Saffari M, et al.
Iranian Journal of Pharmaceutical Research : IJPR, 12, 3-3 (2013)
Nonequilibrium adhesion patterns at lipid bilayer junctions
Parthasarathy R, et al.
The Journal of Physical Chemistry B, 108(2), 649-657 (2004)
Qiang Cheng et al.
Nature nanotechnology, 15(4), 313-320 (2020-04-07)
CRISPR-Cas gene editing and messenger RNA-based protein replacement therapy hold tremendous potential to effectively treat disease-causing mutations with diverse cellular origin. However, it is currently impossible to rationally design nanoparticles that selectively target specific tissues. Here, we report a strategy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service