Skip to Content
Merck
All Photos(1)

Key Documents

880680P

Avanti

C16 PEG750 Ceramide

Avanti Research - A Croda Brand 880680P, powder

Synonym(s):

N-palmitoyl-sphingosine-1-{succinyl[methoxy(polyethylene glycol)750]}

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$874.00

$874.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$874.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$874.00


Estimated to ship on26 April 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (880680P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880680P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCC(OC[C@]([H])(NC(CCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC)=O)OOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC.C=C

InChI key

PZQTVIDFPGJTMH-TWVCMLEGSA-N

General description

Ceramide is a sphingolipid metabolite. The ceramides differ in their acyl chain length.[1]

Application

C16 PEG750 Ceramide might be used as a control to test for nonspecific interactions during titrations of DNA-nanotube complexes with polyethylene glycol (PEG)-conjugated lipids.[2]

Biochem/physiol Actions

Therapeutic drugs enclosed in poly-ethylene glycol (PEG) conjugated ceramide serves as liposomal carriers, ensuring a longer circulation period. It exhibits a neutral surface electrostatic charge which prevents the leaking of positively charged encapsulated drug molecule.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (880680P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Webb et al.
Biochimica et biophysica acta, 1372(2), 272-282 (1998-07-24)
Poly(ethylene glycol) (PEG) conjugated lipids have been used to increase the circulation longevity of liposomal carriers encapsulating therapeutic compounds. PEG is typically conjugated to distearoylphosphatidylethanolamine (DSPE) via a carbamate linkage that results in a net negative charge on the phosphate
Prakrit V Jena et al.
ACS nano, 11(11), 10689-10703 (2017-09-13)
Lipid accumulation within the lumen of endolysosomal vesicles is observed in various pathologies including atherosclerosis, liver disease, neurological disorders, lysosomal storage disorders, and cancer. Current methods cannot measure lipid flux specifically within the lysosomal lumen of live cells. We developed
Jamie L Betker et al.
Journal of pharmaceutical sciences (2020-02-01)
Nanoparticle-mediated drug delivery has long utilized PEGylation as a mechanism for reducing uptake by the reticuloendothelial system and extending circulation lifetimes. However, studies over the past 2 decades have established that immune responses to PEG can promote clearance on repeat injection
Michal Levy et al.
IUBMB life, 62(5), 347-356 (2010-03-12)
In mammals, ceramide, a key intermediate in sphingolipid metabolism and an important signaling molecule, is synthesized by a family of six ceramide synthases (CerS), each of which synthesizes ceramides with distinct acyl chain lengths. There are a number of common

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service