Skip to Content
Merck
All Photos(1)

Key Documents

880344P

Avanti

OChemsPC

Avanti Research - A Croda Brand 880344P, powder

Synonym(s):

1-oleoyl-2-cholesterylhemisuccinoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$468.00

$468.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$468.00

About This Item

Empirical Formula (Hill Notation):
C57H100NO10P
CAS Number:
Molecular Weight:
990.38
UNSPSC Code:
51191904
NACRES:
NA.25

$468.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (880344P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880344P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OCC[N+](C)(C)C)OC[C@H](OC(CCC(OC1CC[C@]2(C)C(C1)=CCC3C2CC[C@]4(C)C3CC[C@H]4[C@@H](C)CCCC(C)C)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C57H100NO10P/c1-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-28-53(59)64-42-48(43-66-69(62,63)65-40-39-58(7,8)9)68-55(61)34-33-54(60)67-47-35-37-56(5)46(41-47)29-30-49-51-32-31-50(45(4)27-25-26-44(2)3)57(51,6)38-36-52(49)56/h17-18,29,44-45,47-52H,10-16

InChI key

YYWBZBZOTINVNZ-OCTSRPJISA-N

Application

OChemsPC might be used to study its pharmacokinetic and biodistribution patterns in comparison with phospholipid-cholesterol liposomes.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (880344P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A robust and quantitative method for tracking liposome contents after intravenous administration
Kohli AG, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 176, 86-93 (2014)
Zhaohua Huang et al.
Angewandte Chemie (International ed. in English), 48(23), 4146-4149 (2009-05-09)
Extreme makeover of cholesterol: Cholesterol exchange is a major reason for the instability of liposomes in blood. The formation of a covalent hybrid between cholesterol and glycerophosphocholine preserves the bilayer-stabilizing effect of free cholesterol but prevents its transfer from the
Maryam Iman et al.
International journal of pharmaceutics, 408(1-2), 163-172 (2011-02-01)
1,2-Di-stigma-steryl-hemi-succinoyl-sn-glycero-3-phosphocholine (DSHemsPC) is a new lipid in which two molecules of stigmasterol (an inexpensive plant sterol) are covalently linked via a succinic acid to glycerophosphocholine. Since amphotericin B (AmB) interacts with sterols, we postulated that DSHemsPC could be used in
F Foglia et al.
Langmuir : the ACS journal of surfaces and colloids, 27(13), 8275-8281 (2011-06-04)
Langmuir isotherm, neutron reflectivity, and small angle neutron scattering studies have been conducted to characterize the monolayers and vesicular bilayers formed by a novel chimeric phospholipid, ChemPPC, that incorporates a cholesteryl moeity and a C-16 aliphatic chain, each covalently linked
Zhaohua Huang et al.
Journal of the American Chemical Society, 130(46), 15702-15712 (2008-10-28)
We synthesized a family of sterol-modified glycerophospholipids (SML) in which the sn-1 or sn-2 position is covalently attached to cholesterol and the alternative position contains an aliphatic chain. The SML were used to explore how anchoring cholesterol to a phospholipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service