Skip to Content
Merck
All Photos(1)

Documents

880210P

Avanti

14:0 PEG5000 PE

Avanti Research - A Croda Brand 880210P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (880210P-25mg)
pkg of 1 × 500 mg (880210P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880210P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC([(OCH2CH2)113OCH3])=O)=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[NH4+]

General description

14:0 PEG5000 PE, also known as 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000], (DMPE-PEG5000) is a polyethylene glycol (PEG)-modified lipid. Insertion of PEG-lipid conjugates into lipid bilayers decreases the rate of protein binding on the complex surface and thus, aids in formation of stabilized liposomes.

Application

14:0 PEG5000 PE or 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000], (DMPE-PEG5000) has been used to modify 1,4-dihydropyridine amphiphile DHP-12 complexes with DNA. It has also been used in preparation of polyethylene glycol (PEG)-ylated liposomes (PL).

Packaging

20 mL Clear Glass Screw Cap Vial (880210P-500mg)
5 mL Amber Glass Screw Cap Vial (880210P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zanna Hyvönen et al.
Journal of controlled release : official journal of the Controlled Release Society, 99(1), 177-190 (2004-09-03)
Complexes of amphiphilic cationic 1,4-dihydropyridines with DNA (lipoplexes) can be used for nonviral gene delivery. In order to achieve serum-resistant transfection system, DOPE and PEG-lipid conjugates were used to modify 1,4-dihydropyridine amphiphile DHP-12 complexes with DNA. The ability to bind
Renee R Hood et al.
Pharmaceutical research, 30(6), 1597-1607 (2013-02-07)
A microfluidic hydrodynamic flow focusing technique enabling the formation of small and nearly monodisperse liposomes is investigated for continuous-flow synthesis of poly(ethylene glycol) (PEG)-modified and PEG-folate-functionalized liposomes for targeted drug delivery. Controlled laminar flow in thermoplastic microfluidic devices facilitated liposome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service