Skip to Content
Merck
All Photos(1)

Key Documents

880121P

Avanti

DSPE-PEG(2000) Succinyl

Avanti Research - A Croda Brand 880121P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[succinyl(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$315.00

$315.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$315.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$315.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (880121P-10mg)
pkg of 1 × 100 mg (880121P-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880121P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\N([H])C(CCC(O)=O)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=

General description

DSPE-PEG(2000) Succinyl is a functionalized polyethylene glycol (PEG) lipid conjugate.

Application

DSPE-PEG(2000) Succinyl has been used to create a lipid monolayer for the synthesis of nanoparticles encapsulating curcumin.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (880121P-100mg)
5 mL Amber Glass Screw Cap Vial (880121P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lipid-polymer nanoparticles encapsulating curcumin for modulating the vascular deposition of breast cancer cells
Palange AL, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 10(5), e991-e1002 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service