Skip to Content
Merck
All Photos(1)

Key Documents

878130C

Avanti

C16-18:1 PE

Avanti Research - A Croda Brand 878130C

Synonym(s):

1-hexadecyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$349.00

$349.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$349.00

About This Item

Empirical Formula (Hill Notation):
C39H78NO7P
CAS Number:
Molecular Weight:
704.01
UNSPSC Code:
12352211
NACRES:
NA.25

$349.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mL (878130C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878130C

concentration

5 mg/mL (878130C-5mg)

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COCCCCCCCCCCCCCCCC

InChI

1S/C39H78NO7P/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-39(41)47-38(37-46-48(42,43)45-35-33-40)36-44-34-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2/h17,19,38H,3-16,18,20-37,40H2,1-2H3,(H,42,43)/b19-17-/t38-/m1/s1

InChI key

JJGMGPGKWYLISM-NPBIGWJUSA-N

Application

C16-18:1 PE or 1-hexadecyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine has been used as a lipid standard in liquid chromatography with tandem mass spectrometry (LC-MS-MS)[1] and LC-20AD prominence high performance liquid chromatography system to quantify glycerophosphoethanolamines.[2]

Packaging

5 mL Clear Glass Sealed Ampule (878130C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Kai Xie et al.
Food chemistry, 281, 251-260 (2019-01-20)
Lipid hydrolysis and oxidation occurred in Argopecten irradians adductor muscle during hot air drying. Using an in vivo imaging system, we found that antioxidants of bamboo leaves (AOB) could diffuse into the adductor muscle upon marinating. Both tea polyphenols (TP)
Ernst R Werner et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(14), 7792-7798 (2020-03-27)
A significant fraction of the glycerophospholipids in the human body is composed of plasmalogens, particularly in the brain, cardiac, and immune cell membranes. A decline in these lipids has been observed in such diseases as Alzheimer's and chronic obstructive pulmonary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service