Skip to Content
Merck
All Photos(1)

Key Documents

878123C

Avanti

C16-20:5 PC

Avanti Research - A Croda Brand 878123C

Synonym(s):

1-O-hexadecyl-2-(5Z,8Z,11Z,14Z,17Z-eicosapentaenoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$875.00

$875.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$875.00

About This Item

Empirical Formula (Hill Notation):
C44H80NO7P
CAS Number:
Molecular Weight:
766.08
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$875.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mL (878123C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878123C

concentration

5 mg/mL (878123C-5mg)

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C44H80NO7P/c1-6-8-10-12-14-16-18-20-22-23-24-25-27-29-31-33-35-37-44(46)52-43(42-51-53(47,48)50-40-38-45(3,4)5)41-49-39-36-34-32-30-28-26-21-19-17-15-13-11-9-7-2/h8,10,14,16,20,22,24-25,29,31,43H,6-7,9,11-13,15,17-19,21,23,26-28,30,32-42H2,1-5H3/b10-8-,16-14-,22-20-,25-24-,31-29-/t43-/m1/s1

InChI key

YHSSHPWVFALMBA-IARQOZKNSA-N

General description

Phophotidylcholine (PC) is a glycerophospholipid and is synthesized by the CDP-choline pathway or Kennedy pathway.[1][2] It consists of 40-50% of the total phospholipids in mammalian cells and cell organelles.[2]

Biochem/physiol Actions

Phophotidylcholine (PC) in hepatocytes are involved in the synthesis of very low density lipoprotein (VLDL).[2]

Packaging

5 mL Clear Glass Sealed Ampule (878123C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The critical role of phosphatidylcholine and phosphatidylethanolamine metabolism in health and disease
van der Veen J N, et al.
Biochimica et Biophysica Acta - Biomembranes (2017)
A review on phospholipids and their main applications in drug delivery systems
Li J, et al.
Asian Journal of Pharmaceutical Sciences , 10(2), 81-98 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service