Skip to Content
Merck
All Photos(1)

Key Documents

878112P

Avanti

C16-18:1 PC

Avanti Research - A Croda Brand 878112P, powder

Synonym(s):

1-O-hexadecyl-2-oleoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$270.00

$270.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$270.00

About This Item

Empirical Formula (Hill Notation):
C42H84NO7P
CAS Number:
Molecular Weight:
746.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$270.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (878112P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878112P

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COCCCCCCCCCCCCCCCC)=O

InChI key

SIEDNCDNGMIKST-IYEJTHTFSA-N

General description

C16-18:1 PC, also called as 1-O-hexadecyl-2-oleoyl-sn-glycero-3-phosphocholine (HOPC), is a member of platelet-activating factor (PAF) family of glycerophospholipids.[1] HOPC is a mono-ether PC containing ester-linked 18:1 chain (oleoyl) at the sn-2 position and an ether-linked 16:0 chain (hexadecyl) at the sn-1 position.[2]

Application

C16-18:1 PC or 1-O-hexadecyl-2-oleoyl-sn-glycero-3-phosphocholine may be used in the liposome binding assay.[2] It may also be used as a lipid standard for quantification of glycerophosphocholine by liquid chromatography–mass spectrometry (LC–MS).[3][4]

Biochem/physiol Actions

Platelet-activating factor (PAF), a lipid mediator acts as a pro-inflammatory agent that exhibits variety of biological and pharmacological effects.[5]

Packaging

5 mL Clear Glass Sealed Ampule (878112P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrey B Imbs et al.
PloS one, 14(4), e0215759-e0215759 (2019-04-30)
To expand our knowledge of lipid and fatty acid (FA) biosynthesis in marine cnidarians, polar lipidomes of hydrocorals were studied for the first time and then compared with those of soft corals from tropical and boreal regions. The structure and
Saki Yamaura et al.
Clinica chimica acta; international journal of clinical chemistry, 481, 184-188 (2018-03-20)
Measurement of lipoprotein-associated phospholipase A2 (Lp-PLA2) can be used as an adjunct to traditional cardiovascular risk factors for identifying individuals at higher risk of cardiovascular events. This can be performed by quantification of the protein concentration using an ELISA platform
John J H Shin et al.
Developmental cell, 52(4), 461-476 (2020-01-14)
Phosphoinositides, diacylglycerolpyrophosphate, ceramide-1-phosphate, and phosphatidic acid belong to a unique class of membrane signaling lipids that contain phosphomonoesters in their headgroups having pKa values in the physiological range. The phosphomonoester headgroup of phosphatidic acid enables this lipid to act as
Scott D Ryan et al.
Journal of lipid research, 49(10), 2250-2258 (2008-06-14)
The platelet-activating factor (PAF) family of glycerophospholipids accumulates in damaged brain tissue following injury. Little is known about the role of individual isoforms in regulating neuronal survival. Here, we compared the neurotoxic and neuroprotective activities of 1-O-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine (C16-PAF) and 1-O-octadecyl-2-acetyl-sn-glycero-3-phosphocholine
Ernst R Werner et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(14), 7792-7798 (2020-03-27)
A significant fraction of the glycerophospholipids in the human body is composed of plasmalogens, particularly in the brain, cardiac, and immune cell membranes. A decline in these lipids has been observed in such diseases as Alzheimer's and chronic obstructive pulmonary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service