Skip to Content
Merck
All Photos(1)

Key Documents

870743P

Avanti

16:1 (n7) Coenzyme A

Avanti Research - A Croda Brand 870743P, powder

Synonym(s):

palmitoleoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$733.00

$733.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$733.00

About This Item

Empirical Formula (Hill Notation):
C37H73N10O17P3S
CAS Number:
Molecular Weight:
1055.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$733.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870743P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870743P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCC/C=C\CCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C37H64N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-28(46)65-21-20-39-27(45)18-19-40-35(49)32(48)37(2,3)23-58-64(55,56)61-63(53,54)57-22-26-31(60-62(50,51)52)30(47)36(59-26)44-25-43-29-33(38)41-24-42-34(29)44;;;/h9-10,24-26,30-32,36,47-48H,4-8,1

InChI key

BZLFGTFLKXOWRJ-SZUAKDMZSA-N

General description

16:1(n7) Coenzyme A or palmitoleoyl Coenzyme A is a derivative of palmitoleic acid. It is a key monounsaturated fatty acid present in cutaneous lipids. It is produced by the insertion of the first cis-double bond at the Δ9-position of palmitoleic acid by Δ9-desaturase.[1]

Biochem/physiol Actions

16:1 (n7) Coenzyme A or palmitoleoyl Coenzyme A has been used as a substrate for 4′-phosphopantetheinyl transferase or Sfp transferase from Bacillus subtilis to produce palmitoleoyl- acyl carrier protein (ACP).[2]

Packaging

5 mL Amber Glass Screw Cap Vial (870743P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harini Sampath et al.
The Journal of biological chemistry, 289(5), 2482-2488 (2013-12-21)
The skin is the single largest organ in humans, serving as a major barrier to infection, water loss, and abrasion. The functional diversity of skin requires the synthesis of large amounts of lipids, such as triglycerides, wax esters, squalene, ceramides
Jeong-Won Nam et al.
The Plant cell, 32(4), 820-832 (2020-02-16)
Acyl carrier proteins (ACPs) are the scaffolds for fatty acid biosynthesis in living systems, rendering them essential to a comprehensive understanding of lipid metabolism. However, accurate quantitative methods to assess individual acyl-ACPs do not exist. We developed a robust method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service