Skip to Content
Merck
All Photos(1)

Key Documents

870735P

Avanti

14:0 Ether Coenzyme A

Avanti Research - A Croda Brand 870735P, powder

Synonym(s):

tetradecyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$733.00

$733.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$733.00

About This Item

Empirical Formula (Hill Notation):
C35H73N10O16P3S
CAS Number:
Molecular Weight:
1015.00
UNSPSC Code:
12352211
NACRES:
NA.25

$733.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870735P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870735P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSCCCCCCCCCCCCCC)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI key

JDSLZFDMZLUDIT-PVWOJWAGSA-N

General description

14:0 Ether Coenzyme A, also known as tetradecyl coenzyme A (CoA), is a long-chain acyl-CoA ester and a thioether analog of myristoyl coenzyme A. Long-chain acyl-CoA esters play an important role in a plethora of cellular functions. These lipids are involved in regulation of fatty acid synthesis, enzyme systems, vesicle trafficking, ion channels and ion pumps.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870735P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Elholm et al.
Journal of lipid research, 41(4), 538-545 (2000-04-01)
A detailed analysis of the subcellular distribution of acyl-CoA esters in rat liver revealed that significant amounts of long-chain acyl-CoA esters are present in highly purified nuclei. No contamination of microsomal or mitochondrial marker enzymes was detectable in the nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service