Skip to Content
Merck
All Photos(1)

Key Documents

870602C

Avanti

PGPC

Avanti Research - A Croda Brand 870602C

Synonym(s):

1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$252.00
5 MG
$809.00

$252.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$252.00
5 MG
$809.00

About This Item

Empirical Formula (Hill Notation):
C29H56NO10P
CAS Number:
Molecular Weight:
609.73
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$252.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mL (870602C-1mg)
pkg of 1 × 1 mL (870602C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870602C

concentration

1 mg/mL (870602C-1mg)
5 mg/mL (870602C-5mg)

lipid type

phospholipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCC(O)=O)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI key

CDZVJFRXJAUXPP-AREMUKBSSA-N

Application

PGPC (1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine) may be used as a standard in mass spectrometry analysis of phospholipids and oxidized phospholipids.[1][2]

Biochem/physiol Actions

1-palmitoyl-2-glutaroyl-sn-glycero-phosphocholine (PGPC) is a truncated oxidized phospholipid (Tr-OxPL).[2] It elicits endothelial barrier disruption.[2] PGPC exhibits proinflammatory properties and increases the production of protective enzyme such as heme oxygenase 1. It acts as a precursor for the synthesis of 1-palmitoyl-2-glutaroyl-sn-glycero-3-phosphoethanolamine (PGPE). PGPC stimulates apoptotic signaling in vascular smooth muscle cells (VSMCs).[3]

Packaging

5 mL Clear Glass Sealed Ampule (870602C-1mg)
5 mL Clear Glass Sealed Ampule (870602C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pratap Karki et al.
PloS one, 13(11), e0206251-e0206251 (2018-11-13)
Particulate matter (PM) air pollution is a global environmental health problem contributing to more severe lung inflammation and injury. However, the molecular and cellular mechanisms of PM-induced exacerbation of lung barrier dysfunction and injury are not well understood. In the
Corinne M Spickett et al.
Free radical biology & medicine, 51(12), 2133-2149 (2011-10-11)
Phospholipids are complex and varied biomolecules that are susceptible to lipid peroxidation after attack by free radicals or electrophilic oxidants and can yield a large number of different oxidation products. There are many available methods for detecting phospholipid oxidation products
Alexandra Moumtzi et al.
Journal of lipid research, 48(3), 565-582 (2006-12-01)
Lipid oxidation is now thought to be an initiating and sustaining event in atherogenesis. Oxidatively fragmented phospholipids, namely 1-palmitoyl-2-glutaroyl-sn-glycero-3-phosphocholine (PGPC) and 1-palmitoyl-2-(5-oxovaleroyl)-sn-glycero-3-phosphocholine (POVPC), present in minimally modified LDL and atherosclerotic lesions, have been reported to elicit a wide range of
Marco Di Gioia et al.
Nature immunology, 21(1), 42-53 (2019-11-27)
Pathogen-associated molecular patterns (PAMPs) have the capacity to couple inflammatory gene expression to changes in macrophage metabolism, both of which influence subsequent inflammatory activities. Similar to their microbial counterparts, several self-encoded damage-associated molecular patterns (DAMPs) induce inflammatory gene expression. However
Mario Brameshuber et al.
Biophysical journal, 110(1), 205-213 (2016-01-09)
We previously developed a single-molecule microscopy method termed TOCCSL (thinning out clusters while conserving stoichiometry of labeling), which allows for direct imaging of stable nanoscopic platforms with raft-like properties diffusing in the plasma membrane. As a consensus raft marker, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service