Skip to Content
Merck
All Photos(1)

Key Documents

870245P

Avanti

16:0 Glutaryl PE

Avanti Research - A Croda Brand 870245P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(glutaryl) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$301.00

$301.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$301.00

About This Item

Empirical Formula (Hill Notation):
C42H79NO11PNa
CAS Number:
Molecular Weight:
828.04
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$301.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (870245P-25mg)
pkg of 1 × 500 mg (870245P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870245P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCC(O)=O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C42H80NO11P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-32-41(47)51-36-38(37-53-55(49,50)52-35-34-43-39(44)30-29-31-40(45)46)54-42(48)33-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h38H,3-37H2,1-2H3,(H,43,44)(H,45,46)(H,49,50);/q;+1/p-1/t38-;/m1./s1

InChI key

ZDVPYKPXKPQSMT-XVYLPRMCSA-M

General description

16:0 Glutaryl PE, also known as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(glutaryl) (N-glutaryl-DPPE) is a head group modified functionalized lipid. Functionalized lipids function as reporter molecules.[1]

Application

16:0 Glutaryl PE is suitable for liposome preparation.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (870245P-500mg)
5 mL Amber Glass Screw Cap Vial (870245P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cornelia A Hermann et al.
Analytical and bioanalytical chemistry (2020-02-20)
Magnetized liposome (magnetosomes) labels can overcome diffusion limitations in bioassays through fast and easy magnetic attraction. Our aim therefore was to advance the understanding of factors influencing their synthesis focusing on encapsulation strategies and synthesis parameters. Magnetosome synthesis is governed
Baksun Kim et al.
Journal of controlled release : official journal of the Controlled Release Society, 322, 530-541 (2020-04-11)
Here, we report rationally engineered peptide-targeted liposomal doxorubicin nanoparticles that have an enhanced selectivity for HER2-positive breast tumor cells with high purity, reproducibility, and precision in controlling stoichiometry of targeting peptides. To increase HER2-positive tumor cell selective drug delivery, we
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service