Skip to Content
Merck
All Photos(1)

Key Documents

870205P

Avanti

16:0 PDP PE

Avanti Research - A Croda Brand 870205P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[3-(2-pyridyldithio)propionate] (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$1,330.00

$1,330.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$1,330.00

About This Item

Empirical Formula (Hill Notation):
C45H80N2O9PNaS2
CAS Number:
Molecular Weight:
911.22
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,330.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (870205P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870205P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCSSC1=CC=CC=N1)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C45H81N2O9PS2.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-32-44(49)53-39-41(56-45(50)33-28-26-24-22-20-18-16-14-12-10-8-6-4-2)40-55-57(51,52)54-37-36-46-42(48)34-38-58-59-43-31-29-30-35-47-43;/h29-31,35,41H,3-28,32-34,36-40H2,1-2H3,(H,46,48)(H,51,52);/q;+1/p-1/t41-;/m1./s1

InChI key

IUSHQKJFHVODPF-QUEDAWNRSA-M

General description

16:0 PDP PE, also referred as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[3-(2-pyridyldithio) propionate, belongs to a class of head group modified functionalized lipids. Functionalized lipids are primarily used as reporter molecules.[1]

Application

16:0 PDP PE has been used in preparation of rhodamine high-density lipoprotein nanoparticle (Rh-HDL NP) synthesis.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (870205P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pathways for modulating exosome lipids identified by high-density lipoprotein-like nanoparticle binding to scavenger receptor type B-1
Angeloni NL, et al.
Scientific Reports, 6(1) (2016)
Pathways for modulating exosome lipids identified by high-density lipoprotein-like nanoparticle binding to scavenger receptor type B-1
Angeloni NL, et al.
Scientific reports, 6(1) (2016)
Functionalized lipids and surfactants for specific applications
Kepczynski M and Rog T
Biochimica et Biophysica Acta - Biomembranes, 1858(10), 2362-2379 (2016)
Nicholas L Angeloni et al.
Scientific reports, 6, 22915-22915 (2016-03-12)
Exosomes are produced by cells to mediate intercellular communication, and have been shown to perpetuate diseases, including cancer. New tools are needed to understand exosome biology, detect exosomes from specific cell types in complex biological media, and to modify exosomes.
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service