Skip to Content
Merck
All Photos(1)

Key Documents

870125P

Avanti

16:0 Caproylamine PE

Avanti Research - A Croda Brand 870125P, powder

Synonym(s):

1,2-Dipalmitoyl-sn-Glycero-3-Phosphoethanolamine-N-(hexanoylamine)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$292.00

$292.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$292.00

About This Item

Empirical Formula (Hill Notation):
C43H85N2O9P
CAS Number:
Molecular Weight:
805.12
UNSPSC Code:
12352211
NACRES:
NA.25

$292.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (870125P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870125P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCCC[NH3+])=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C43H85N2O9P/c1-3-5-7-9-11-13-15-17-19-21-23-25-29-33-42(47)51-38-40(39-53-55(49,50)52-37-36-45-41(46)32-28-27-31-35-44)54-43(48)34-30-26-24-22-20-18-16-14-12-10-8-6-4-2/h40H,3-39,44H2,1-2H3,(H,45,46)(H,49,50)/t40-/m1/s1

InChI key

IVLBQYFFAUWDCA-RRHRGVEJSA-N

General description

16:0 Caproylamine PE is a head group modified functionalized lipid. Caproylamine is a lipid containing free amino group on the lipid head, providing the flexibility to bind biomolecules that are important for drug delivery or target tissue specificity.[1]

Application

16:0 Caproylamine PE is suitable for the synthesis of dibenzocyclooctyne (DBCO)-oligo(ethylene glycol)-lipid (φ-OEG-lipid) for producing supported lipid bilayer microarray.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (870125P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Shen et al.
ACS chemical biology, 9(8), 1877-1884 (2014-06-21)
The binding of lectins to glycan receptors on the host cell surface is a key step contributing to the virulence and species specificity of most viruses. This is exemplified by the viral protein hemagglutinin (HA) of the influenza A virus
Luís F Neves et al.
Journal of microencapsulation, 33(4), 391-399 (2016-08-18)
Drug delivery to corneal epithelial cells is challenging due to the intrinsic mechanisms that protect the eye. Here, we report a novel liposomal formulation to encapsulate and deliver a short sequence peptide into human corneal epithelial cells (hTCEpi). Using a
Lucas H Hofmeister et al.
ACS nano, 9(4), 4435-4446 (2015-03-15)
In regions of the circulation where vessels are straight and unbranched, blood flow is laminar and unidirectional. In contrast, at sites of curvature, branch points, and regions distal to stenoses, blood flow becomes disturbed. Atherosclerosis preferentially develops in these regions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service