Skip to Content
Merck
All Photos(1)

Key Documents

860848P

Avanti

CER5-2′R(d9)

Avanti Research - A Croda Brand 860848P, powder

Synonym(s):

N-(2′-(R)-hydroxypalmitoyl(d9)) D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$769.00

$769.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$769.00

About This Item

Empirical Formula (Hill Notation):
C34H58D9NO4
CAS Number:
Molecular Weight:
562.96
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

$769.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860848P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860848P

lipid type

sphingolipids
deuterated lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@H](O)CCCCCCCCCCC([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])=O)CO

InChI key

YWZGQPJRQXHVQX-FMUVJMMOSA-N

General description

CER5-2′R(d9), also known as N-(2′-(R)-hydroxypalmitoyl(d9)) D-erythro-sphingosine has been used as a deuterium-labeled ceramide standard in liquid chromatography-mass spectrometry (LC-MS)/MS for qualitative analysis of stratum corneum (SC) ceramides in humans and mice.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860848P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Momoko Kawana et al.
Journal of lipid research, 61(6), 884-895 (2020-04-09)
Ceramides are the predominant lipids in the stratum corneum (SC) and are crucial components for normal skin barrier function. Although the composition of various ceramide classes in the human SC has been reported, that in mice is still unknown, despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service