Skip to Content
Merck
All Photos(1)

Key Documents

860691P

Avanti

C24:1 Ceramide (d18:2(4E,8Z)/24:1(15Z))

Avanti Research - A Croda Brand 860691P, powder

Synonym(s):

N-nervonoyl-D-erythro-sphingosine (d18:2(4E,8Z) base)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$585.00

$585.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$585.00

About This Item

Empirical Formula (Hill Notation):
C42H79NO3
CAS Number:
Molecular Weight:
646.08
UNSPSC Code:
12352211
NACRES:
NA.25

$585.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860691P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860691P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)([H])[C@]([H])(O)/C=C/CC/C=C\CCCCCCCCC

InChI

1S/C42H79NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-42(46)43-40(39-44)41(45)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h17-18,27,29,35,37,40-41,44-45H,3-16,19-26,28,30-34,36,38-39H2,1-2H3,(H,43,46)/b18-17-,29-27-,37-35+/t40-,41+/m0

InChI key

RWMLFFULUMHLPQ-WGCGRUQKSA-N

General description

C24:1 Ceramide (d18:2(4E,8Z)/24:1(15Z)) or N-nervonoyl-D-erythro-sphingosine (d18:2(4E,8Z) base), is a synthetic ceramide derived from sphingosine with nervonic acid.

Biochem/physiol Actions

C24:1 Ceramide in a stable isotope form is suitable for a newly described validated liquid chromatography-tandem mass spectrometry methodology for their clinical-scaled measurement.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860691P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimple Kauhanen et al.
Analytical and bioanalytical chemistry, 408(13), 3475-3483 (2016-02-29)
Monitoring the levels of the ceramides (Cer) d18:1/16:0, Cer d18:1/18:0, Cer d18:1/24:0, and Cer d18:1/24:1 and ratios thereof in human plasma empowers the prediction of fatal outcome of coronary artery disease (CAD). We describe a validated liquid chromatography-tandem mass spectrometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service