Skip to Content
Merck
All Photos(1)

Key Documents

860679L

Avanti

C24:1 Ceramide-d7 (d18:1-d7/24:1)

Avanti Research - A Croda Brand 860679L-1MG860746L-1MG

Synonym(s):

N-nervonoyl-D-erythro-sphingosine (d7)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,910.00

$1,910.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$1,910.00

About This Item

Empirical Formula (Hill Notation):
C42H74D7NO3
CAS Number:
Molecular Weight:
655.14
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.12

$1,910.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

solution

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand 860679L-1MG
860746L-1MG

concentration

1 mg/mL

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Glass Sealed Ampule (860679L-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hady Razak Hady et al.
Journal of clinical medicine, 8(12) (2019-12-18)
The liver plays a central role in the glucose and lipid metabolism. Studies performed on animal models have shown an important role of lipid accumulation in the induction of insulin resistance. We sought to explain whether in obese humans, the
Tejia Zhang et al.
Scientific reports, 9(1), 19939-19939 (2019-12-29)
Extensive characterisations of the zebrafish genome and proteome have established a foundation for the use of the zebrafish as a model organism; however, characterisation of the zebrafish lipidome has not been as comprehensive. In an effort to expand current knowledge

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service