Skip to Content
Merck
All Photos(1)

Key Documents

860671P

Avanti

C16 Ceramide (d14:1/16:0)

Avanti Research - A Croda Brand, powder

Synonym(s):

N-palmitoyl-D-erythro-sphingosine (C14 base)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$2,340.00

$2,340.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$2,340.00

About This Item

Empirical Formula (Hill Notation):
C30H59NO3
CAS Number:
Molecular Weight:
481.79
UNSPSC Code:
12352100
NACRES:
NA.25

$2,340.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

1 ea of 10 mg (860671P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

deuterated lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCC

InChI

1S/C30H59NO3/c1-3-5-7-9-11-13-14-15-16-18-20-22-24-26-30(34)31-28(27-32)29(33)25-23-21-19-17-12-10-8-6-4-2/h23,25,28-29,32-33H,3-22,24,26-27H2,1-2H3,(H,31,34)/b25-23+/t28-,29+/m0/s1

InChI key

OCXQIBPNDRJADM-VARSQMIESA-N

General description

Ceramide is a sphingolipid metabolite.[1] It is composed of sphingosine base linked via amide linkage to a fatty acid.[2]

Biochem/physiol Actions

Ceramide plays a role in cell signaling processes, such as cell cycle arrest and promotes apoptosis. It is also involved in signal transduction processes and insulin resistance.[3] Ceramide also plays a role in cell proliferation, differentiation and inflammatory responses.[1]

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramides and skin function
Coderch L, et al.
American Journal of Clinical Dermatology, 4(2), 107-129 (2003)
Ceramide and ceramide 1-phosphate in health and disease
Arana L, et al.
Lipids in Health and Disease, 9(1), 15-15 (2010)
Ceramide 1-phosphate/ceramide, a switch between life and death
Gomez-Munoz A
Biochimica et Biophysica Acta - Biomembranes, 1758(12), 2049-2056 (2006)
Mutay Aslan et al.
Nutrition and cancer, 1-13 (2020-04-15)
We aimed to investigate the impact of thymoquinone (TQ), on sphingolipid metabolites, ER stress and apoptotic pathways in MCF-7 and HepG2 cancer cells. Antiproliferative effect was exerted in cancer cells via TQ incubation at different doses and durations. Cell viability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service