Skip to Content
Merck
All Photos(1)

Key Documents

860657P

Avanti

sphingosine-d7

Avanti Research - A Croda Brand 860657P, powder

Synonym(s):

D-erythro-sphingosine-d7

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$484.00
5 MG
$2,110.00

$484.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$484.00
5 MG
$2,110.00

About This Item

Empirical Formula (Hill Notation):
C18H30D7NO2
CAS Number:
Molecular Weight:
306.54
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$484.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860657P-1mg)
pkg of 1 × 5 mg (860657P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860657P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@@](O)([H])/C=C/CCCCCCCCCCC(C(C([2H])([2H])[2H])([2H])[2H])([2H])[2H]

InChI key

WWUZIQQURGPMPG-AVMMVCTPSA-N

General description

Sphingosine-d7 is a deuterated derivative of sphingosine. Sphingosine is an important bioactive sphingolipid metabolite.[1] It is a 18-carbon amino alcohol derived from sphingomyelin. Sphingosine contains an unsaturated hydrocarbon chain.[2]

Application

Sphingosine-d7 has been used as an internal standard in liquid chromatography–tandem mass spectrometry for quantitative analysis of sphingolipids in biological samples.[3][4][5]

Biochem/physiol Actions

Sphingosine negatively regulates cell proliferation and induces apoptosis.[1] It has an ability to regulate the activities of phospholipases, protein kinases, ion channels, cannabinoid receptor type 1 (CB-1) receptors and steroidogenic factor 1 (SF-1) receptors.[2] Sphingosine acts as a precursor for ceramide synthesis.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (860657P-1mg)
5 mL Amber Glass Screw Cap Vial (860657P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

the
Lipid Mediators and Their Metabolism in the Brain (2011)
Stephanie Schwalm et al.
The American journal of pathology, 187(11), 2413-2429 (2017-08-16)
Kidney fibrosis is a hallmark of chronic kidney disease and leads to extracellular matrix accumulation, organ scarring, and loss of kidney function. In this study, we investigated the role of sphingosine kinase-2 (SPHK2) on the progression of tubular fibrosis by
Henning Carstens et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 53(6), 1015-1028 (2019-12-20)
Pulmonary infections with Pseudomonas aeruginosa (P. aeruginosa) or Staphylococcus aureus (S. aureus) are of utmost clinical relevance in patients with cystic fibrosis, chronic obstructive pulmonary disease, after trauma and burn, upon ventilation or in immuno-compromised patients. Many P. aeruginosa and
Iulia Zoicas et al.
Cells, 9(5) (2020-05-24)
Human and murine studies identified the lysosomal enzyme acid sphingomyelinase (ASM) as a target for antidepressant therapy and revealed its role in the pathophysiology of major depression. In this study, we generated a mouse model with overexpression of Asm (Asm-tgfb)
Irina Alecu et al.
Journal of lipid research, 58(1), 60-71 (2016-11-23)
The 1-deoxysphingolipids (1-deoxySLs) are atypical sphingolipids (SLs) that are formed when serine palmitoyltransferase condenses palmitoyl-CoA with alanine instead of serine during SL synthesis. The 1-deoxySLs are toxic to neurons and pancreatic β-cells. Pathologically elevated 1-deoxySLs cause the inherited neuropathy, hereditary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service