Skip to Content
Merck
All Photos(2)

Key Documents

860645P

Avanti

C8 Ceramide (d17:1/8:0)

Avanti Research - A Croda Brand 860645P, powder

Synonym(s):

N-octanoyl-D-erythro-sphingosine (C17 base)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$1,240.00

$1,240.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$1,240.00

About This Item

Empirical Formula (Hill Notation):
C25H49NO3
CAS Number:
Molecular Weight:
411.66
UNSPSC Code:
12352211
NACRES:
NA.25

$1,240.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (860645P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860645P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCC

InChI

1S/C25H49NO3/c1-3-5-7-9-10-11-12-13-14-15-17-18-20-24(28)23(22-27)26-25(29)21-19-16-8-6-4-2/h18,20,23-24,27-28H,3-17,19,21-22H2,1-2H3,(H,26,29)/b20-18+/t23-,24+/m0/s1

InChI key

YUKGJYMYMCFXEF-VYQUXUJXSA-N

General description

C8-ceramide or N-octanoyl-D-erythro-sphingosine (C17 base) is a synthetic cell-permeable sphingolipid[1]. It is a combination of C17 analog of sphingosine and octanoic acid. Ceramides are the major lipid component in the outermost layer of the skin, the stratum corneum.[2] C8 Ceramide plays a vital role in inducing apoptotic cell death.[3]

Application

C8 Ceramide (d17:1/8:0) has been used to induce myriocin-treated malignant melanoma cells to increase endogenous ceramide levels.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860645P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Witty et al.
Endocrinology, 137(12), 5269-5277 (1996-12-01)
Recent studies have demonstrated that ovarian follicle atresia occurs extensively before follicle selection into the avian preovulatory hierarchy, and that this process is mediated via granulosa cell apoptosis. Subsequent to follicle selection, granulosa cells are inherently resistant to apoptosis, and
Y-S Lee et al.
Cell proliferation, 44(4), 320-329 (2011-06-08)
Melanoma is the most aggressive form of skin cancer, and it resists chemotherapy. Candidate drugs for effective anti-cancer treatment have been sought from natural resources. Here, we have investigated anti-proliferative activity of myriocin, serine palmitoyltransferase inhibitor, in the de novo
Luisa Coderch et al.
American journal of clinical dermatology, 4(2), 107-129 (2003-01-30)
Ceramides are the major lipid constituent of lamellar sheets present in the intercellular spaces of the stratum corneum. These lamellar sheets are thought to provide the barrier property of the epidermis. It is generally accepted that the intercellular lipid domain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service