Skip to Content
Merck
All Photos(1)

Key Documents

860464P

Avanti

N-C24:1-deoxysphinganine

Avanti Research - A Croda Brand 860464P, powder

Synonym(s):

N-nervonoyl-1-deoxysphinganine (m18:0/24:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$616.00

$616.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$616.00

About This Item

Empirical Formula (Hill Notation):
C42H83NO2
CAS Number:
Molecular Weight:
634.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$616.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860464P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860464P

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI key

XBNZFPFCTSJHRI-FIIAPBGQSA-N

General description

1-deoxysphinganine is an atypical sphingoid base, which lacks a 1-hydroxyl group.[1]

Application

N-C24:1-deoxysphinganine or N-nervonoyl-1-deoxysphinganine (m18:0/24:1) has been used as a standard in the quantitation of  atypical sphingoid bases in biological samples by reverse-phase liquid chromatography coupled to electrospray ionization tandem mass spectrometry (LC-MS/MS).[1]

Biochem/physiol Actions

1-deoxysphinganine might be toxic to cancer cells.[1] It is a potential biomarker for type 2 diabetes.[2] 1-deoxysphinganine acts as a cytotoxic lipid for insulin producing cells.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860464P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard A Zuellig et al.
Diabetes, 63(4), 1326-1339 (2014-01-01)
Irreversible failure of pancreatic β-cells is the main culprit in the pathophysiology of diabetes, a disease that is now a global epidemic. Recently, elevated plasma levels of deoxysphingolipids, including 1-deoxysphinganine, have been identified as a novel biomarker for the disease.
Junliang Wan et al.
Journal of agricultural and food chemistry, 67(46), 12953-12961 (2019-10-23)
Most common sphingolipids are comprised of "typical" sphingoid bases (sphinganine, sphingosine, and structurally related compounds) and are produced via the condensation of l-serine with a fatty acyl-CoA by serine palmitoyltransferase. Some organisms, including mammals, also produce "atypical" sphingoid bases that
Noemi Jiménez-Rojo et al.
Biophysical journal, 107(12), 2850-2859 (2014-12-18)
Ceramides and dihydroceramides are N-acyl derivatives of sphingosine and sphinganine, respectively, which are the major sphingoid-base backbones of mammals. Recent studies have found that mammals, like certain other organisms, also produce 1-deoxy-(dihydro)ceramides (1-deoxyDHCers) that contain sphingoid bases lacking the 1-hydroxyl-
Sarah T Pruett et al.
Journal of lipid research, 49(8), 1621-1639 (2008-05-24)
"Sphingosin" was first described by J. L. W. Thudichum in 1884 and structurally characterized as 2S,3R,4E-2-aminooctadec-4-ene-1,3-diol in 1947 by Herb Carter, who also proposed the designation of "lipides derived from sphingosine as sphingolipides." This category of amino alcohols is now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service