Skip to Content
Merck
All Photos(3)

Key Documents

860063C

Avanti

Milk SM

Sphingomyelin (Milk, Bovine), chloroform

Synonym(s):

N-(tricosanoyl)-sphing-4-enine-1-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$301.00

$301.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$301.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$301.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (860063C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860063C

concentration

10 mg/mL (860063C-25mg)

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](NC(=O)CCCCCCCCCCCCCCCCCCCCCC)[C@H](O)\C=C\CCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C46H93N2O6P/c1-6-8-10-12-14-16-18-20-21-22-23-24-25-26-28-30-32-34-36-38-40-46(50)47-44(43-54-55(51,52)53-42-41-48(3,4)5)45(49)39-37-35-33-31-29-27-19-17-15-13-11-9-7-2/h37,39,44-45,49H,6-36,38,40-43H2,1-5H3,(H-,47,50,51,52)/b39-37+/t44-,45+/m0/s1

InChI key

SXZWBNWTCVLZJN-NMIJJABPSA-N

General description

As a major constituent of cell membranes, sphingomyelin is found at particularly high concentrations in the membranes of nerve cells (in the myelin sheaths) and red blood cells. It was previously thought to have a purely structural role, similar to the function of phosphatidylcholine, through intermolecular interactions mediated by the 2-amide group, the 3-hydroxy group and the 4,5-trans double bond of the sphingoid base.[1] However, it is now appreciated that sphingomyelin has a high affinity for cholesterol and that these two lipids pack tightly into liquid-ordered domains among a liquid-disordered phase to form lipid rafts.[1] These membrane microdomains are thought to function as signaling platforms that regulate the localization and interactions of proteins. But sphingomyelin does not just influence signaling as a component of lipid rafts — it is also a precursor to ceramides and other sphingolipid metabolites that comprise the sphingomyelin cycle or sphingolipid network.[1]
Sphingomyelin and related lipids

Application

Milk SM is suitable:
  • for detecting ceramide in feces[2]
  • for differential scanning calorimetry[3]
  • as a lipid standard[4]

Packaging

5 mL Clear Glass Sealed Ampule (860063C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Jyonouchi et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 44(1), 58-68 (2013-10-15)
Eosinophilic esophagitis (EoE) is an atopic disease characterized by eosinophilic inflammation in which dietary antigens (in particular, milk) play a major role. EoE is most likely a mixed IgE and non-IgE food-mediated reaction in which overexpression of Th2 cytokines, particularly
Neutral ceramidase encoded by the Asah2 gene is essential for the intestinal degradation of sphingolipids
Kono M, et al.
The Journal of biological chemistry, 281(11), 7324-7331 (2006)
Polymer resonators sensors for detection of sphingolipid gel/fluid phase transition and melting temperature measurement
Li Q, et al.
Sensors and actuators A, Physical, 263, 707-717 (2017)
Mari Kono et al.
The Journal of biological chemistry, 281(11), 7324-7331 (2005-12-29)
Complex sphingolipids are abundant as eukaryotic cell membrane components, whereas their metabolites, in particular ceramide, sphingosine, and sphingosine 1-phosphate, are involved in diverse cell signaling processes. In mammals, degradation of ceramide by ceramidase yields sphingosine, which is phosphorylated by the
A relative quantitative positive/negative ion switching method for untargeted lipidomics via high resolution LC-MS/MS from any biological source
Breitkopf S B, et al.
Metabolomics, 13(3), 30-30 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service