Skip to Content
Merck
All Photos(1)

Key Documents

858126C

Avanti

13:0 Lyso PG

1-tridecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1-tridecanoyl-2-hydroxy-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(13:0/0:0); 110709

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$328.00

$328.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$328.00

About This Item

Empirical Formula (Hill Notation):
C19H38NaO9P
CAS Number:
Molecular Weight:
464.46
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$328.00


Available to ship on15 April 2025Details


Request a Bulk Order

Assay

>99% (LPG; may contain up to 10% of the 2-LPG isomer, TLC)

form

liquid

packaging

pkg of 1 × 1 mL (858126C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 858126C

concentration

5 mg/mL (858126C-5mg)

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule (858126C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service