Skip to Content
Merck
All Photos(1)

Key Documents

857410P

Avanti

IG7

Avanti Research - A Croda Brand, powder

Synonym(s):

N-((2S,3R)-3-hydroxy-1-oxo-1-(tridecylamino)butan-2-yl)-5-methoxy-1H-indole-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$302.00
5 MG
$906.00

$302.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$302.00
5 MG
$906.00

About This Item

Empirical Formula (Hill Notation):
C27H43N3O4
CAS Number:
Molecular Weight:
473.65
UNSPSC Code:
12352211
NACRES:
NA.25

$302.00


Estimated to ship on31 May 2025


Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857410P-5mg)
pkg of 1 × 5 mg (857410P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@H](O)[C@H](NC(C(N1)=CC2=C1C=CC(OC)=C2)=O)C(NCCCCCCCCCCCCC)=O

InChI key

RAXJRRQLMVPSON-NLFFAJNJSA-N

General description

IG7 is a ceramide analog.[1]

Biochem/physiol Actions

IG7 is potent in enhancing Fas ligand (FasL) induced apoptosis in colon carcinoma cells SW480 and RKO. IG7 enhances FasL mediated phosphorylation of p38 and janus kinase (JNK). IG7 enhances nuclear factor-kappa B (NF-κB) activation by FasL. IG7 exhibits low toxicity.[1]

Packaging

5ML Glass Amber Screw Cap Vial (857410P-1MG)
5ML Glass Amber Screw Cap Vial (857410P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide mediates FasL-induced caspase 8 activation in colon carcinoma cells to enhance FasL-induced cytotoxicity by tumor-specific cytotoxic T lymphocytes
Coe GL, et al.
Scientific Reports, 6, 30816-30816 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service