Skip to Content
Merck
All Photos(2)

Key Documents

857400P

Avanti

C6-Urea-Ceramide

D-erythro-N-[2-(1,3-dihydroxy-4E-octadecene)]-N′-hexane-urea-sphingosine, powder

Synonym(s):

D-erythro-C6-urea-ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H50N2O3
CAS Number:
Molecular Weight:
426.68
UNSPSC Code:
12352211

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857400P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857400P

shipped in

dry ice

storage temp.

−20°C

General description

C6-urea ceramide is a N-acylsphingosine amidohydrolase 1 (ASAH2) inhibitor.

Biochem/physiol Actions

C6-urea ceramide helps to decrease the viability of human colon cancer cell lines. It enhances apoptosis and autophagic flux.

Packaging

5 mL Amber Glass Screw Cap Vial (857400P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therapeutic implications of bioactive sphingolipids: A focus on colorectal cancer
Camp ER, et al.
Cancer Biology & Therapy, 18(9), 640-650 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service