856709L
Avanti
15:0 Lyso PE-d5
Avanti Research™ - A Croda Brand
Synonym(s):
1-pentadecanoyl-2-hydroxy-sn-glycero(d5)-3-phosphoethanolamine
Select a Size
About This Item
Recommended Products
Assay
>99% (TLC)
form
solution
packaging
package of 1 × 1 mg
manufacturer/tradename
Avanti Research™ - A Croda Brand
concentration
1 mg/mL
shipped in
dry ice
storage temp.
−20°C
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3
Target Organs
Central nervous system, Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F
Flash Point(C)
9.7 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service