Skip to Content
Merck
All Photos(2)

Key Documents

855375P

Avanti

10:0 Lyso PC

1-decanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-decanoyl-sn-glycero-3-phosphocholine; PC(10:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$278.00

$278.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$278.00

About This Item

Empirical Formula (Hill Notation):
C18H38NO7P
CAS Number:
Molecular Weight:
411.47
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$278.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855375P-25mg)
pkg of 1 × 500 mg (855375P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855375P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCC)=O

InChI

1S/C18H38NO7P/c1-5-6-7-8-9-10-11-12-18(21)24-15-17(20)16-26-27(22,23)25-14-13-19(2,3)4/h17,20H,5-16H2,1-4H3/t17-/m1/s1

InChI key

SECPDKKEUKDCPG-QGZVFWFLSA-N

General description

Lyso-phosphatidylcholine (LPC) is a bioactive proinflammatory lipid secreted on pathological response.[1] It has a choline group at its polar head.[2]

Application

10:0 Lyso PC has been used as an internal standard to quantify the most abundant lysophosphatidylcholine (LPC) species found in Trypanosoma cruzi samples.[3]

Biochem/physiol Actions

Lysophosphatidylcholine (LPC) alters various biological functions in different cell-types, such as endothelial cells, smooth muscle cells, monocytes, macrophages and T-cells. It is associated with atherosclerosis and inflammatory diseases. LPC activates many second messengers including, protein kinase C, extracellular-signal-regulated kinases and protein tyrosine kinases.[1]

Packaging

20 mL Clear Glass Screw Cap Vial (855375P-500mg)
5 mL Clear Glass Sealed Ampule (855375P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felipe Gazos-Lopes et al.
PLoS neglected tropical diseases, 8(8), e3077-e3077 (2014-08-08)
Trypanosoma cruzi is the causative agent of the life-threatening Chagas disease, in which increased platelet aggregation related to myocarditis is observed. Platelet-activating factor (PAF) is a potent intercellular lipid mediator and second messenger that exerts its activity through a PAF-specific
Analysis of phospholipid species in rat peritoneal surface layer by liquid chromatography/electrospray ionization ion-trap mass spectrometry
Gao F, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1761(7), 667-676 (2006)
Role of lysophosphatidylcholine (LPC) in atherosclerosis
Matsumoto T, et al.
Current Medicinal Chemistry, 14(30), 3209-3220 (2007)
R E Stafford et al.
Biochemistry, 28(12), 5113-5120 (1989-06-13)
The critical micelle concentration (cmc) of several lysophospholipids and of a lysophospholipid analogue was determined from surface tension measurements using the maximum bubble pressure method and/or 31P NMR. The use of the maximum bubble pressure method has now been extended

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service