Skip to Content
Merck
All Photos(2)

Key Documents

850852C

Avanti

18:1 Dimethyl PE

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N,N-dimethyl, chloroform

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine-N,N-dimethyl; phosphatidyldimethylethanolamine; PDME

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$394.00

$394.00


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$394.00

About This Item

Empirical Formula (Hill Notation):
C43H82NO8P
CAS Number:
Molecular Weight:
772.09
UNSPSC Code:
51191904
NACRES:
NA.25

$394.00


Estimated to ship on19 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850852C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850852C

concentration

10 mg/mL (850852C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)[H])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C43H82NO8P/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-42(45)49-39-41(40-51-53(47,48)50-38-37-44(3)4)52-43(46)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2/h19-22,41H,5-18,23-40H2,1-4H3,(H,47,48)/b21-19-,22-20-/t41-/m1/s1

InChI key

XHPZRQBHFOVLEJ-UNUIOPIBSA-N

Packaging

5 mL Clear Glass Sealed Ampule (850852C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service