Skip to Content
Merck
All Photos(2)

Key Documents

850537P

Avanti

Facade-TEM

3α,7α,12α-tri-((O-β-D-maltopyranosyl)ethyloxy)-cholane, powder

Synonym(s):

FA-4

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$367.00

$367.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$367.00

About This Item

Empirical Formula (Hill Notation):
C66H114O36
CAS Number:
Molecular Weight:
1483.59
UNSPSC Code:
12161902
NACRES:
NA.25

$367.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

mol wt

1483.59 g/mol

packaging

pkg of 1 × 25 mg (850537P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850537P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@@H](OCCO[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]4[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O4)[C@@H](CO)O3)C5)(C)[C@]5([H])C[C@H]2OCCO[C@@H]6[C@@H](O)[C@H](O)[C@@H](O[C@H]7[C@@H](O)[C@H](O)[C@@H](O)[C@H](CO)O7)[C@H](CO)O6)([H])C[C@H](OCCO[C@

General description

FacadeTEM/FA-4 (3α,7α,12α-tri-((O-β-D-maltopyranosyl)ethyloxy)-cholane) is a sterol-based[1]/sugar-based[2] facial amphiphile.[1][2] It carries three sugar attachments on the steroid backbone.[2]

Biochem/physiol Actions

FacadeTEM/FA-4 (3α,7α,12α-tri-((O-β-D-maltopyranosyl)ethyloxy)-cholane) can be used in the preparation of micelles. It may also be used in the stabilization and crystallization of membrane proteins.[2] FA-4 is not effective as a cystic fibrosis transmembrane conductance regulator (CFTR) solubilizer.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (850537P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Facade is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Designing facial amphiphiles for the stabilization of integral membrane proteins.
Qinghai Zhang et al.
Angewandte Chemie (International ed. in English), 46(37), 7023-7025 (2007-08-11)
Sung Chang Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(13), E1203-E1211 (2013-03-13)
Amphiphile selection is a critical step for structural studies of membrane proteins (MPs). We have developed a family of steroid-based facial amphiphiles (FAs) that are structurally distinct from conventional detergents and previously developed FAs. The unique FAs stabilize MPs and
Ellen Hildebrandt et al.
Biochimica et biophysica acta, 1838(11), 2825-2837 (2014-07-30)
Structural knowledge of the cystic fibrosis transmembrane conductance regulator (CFTR) requires developing methods to purify and stabilize this aggregation-prone membrane protein above 1mg/ml. Starting with green fluorescent protein- and epitope-tagged human CFTR produced in mammalian cells known to properly fold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service