Skip to Content
Merck
All Photos(2)

Key Documents

850530O

Avanti

7.7 MAG

Avanti Research - A Croda Brand 850530O

Synonym(s):

1-(7Z-tetradecenoyl)-rac-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$1,210.00
500 MG
$2,600.00
1 G
$4,590.00

$1,210.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$1,210.00
500 MG
$2,600.00
1 G
$4,590.00

About This Item

Empirical Formula (Hill Notation):
C17H32O4
CAS Number:
Molecular Weight:
300.43
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,210.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

oil

mol wt

300.43 g/mol

packaging

pkg of 10 × 0.1 g (with screw cap (850530O-1g))
pkg of 2 × 100 mg (with screw cap (850530O-200mg))
pkg of 5 × 100 mg (with screw cap (850530O-500mg))

manufacturer/tradename

Avanti Research - A Croda Brand 850530O

technique(s)

protein sequencing: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Monoacylglycerols (MAGs) are glycerol esters.[1] 7.7 MAG has a 14 carbon acyl chain[2] and possess a bilayer thickness of 25.8 Å.[3]
This new Lipid detergent may be used for crystallizing membrane proteins by using lipidic mesophases. This method has variously been referred to as the lipidic cubic phase or in meso method. The method has been shown to be quite versatile in that it has been used to solve X-ray crystallographic structures of prokaryotic and eukaryotic proteins, proteins that are monomeric, homo- and hetero-multimeric, chromophore-containing and chromophore-free, and alpha-helical and beta-barrel proteins.

Application

7.7 MAG may be used as a host lipid in MdfA (multidrug transporter protein)–YN1074 (Fab fragment) complex for crystallization by lipidic cubic phase (LCP) method.[4]

Biochem/physiol Actions

Monoacylglycerols (MAGs) are used in food industry as emulsifiers.[1] 7.7 MAG participates in meso crystallization of BtuB (vitamin B12 transporter), an outer membrane protein. It can also help in lysozyme crystal growth. 7.7 MAG also plays a key role in the growth of diffraction-quality crystals.[5] MAGs holds high significance in pharmaceutical and food industry.[6]

Packaging

5 mL PTFE Vial with Screw Cap (850530O-1g)
5 mL PTFE Vial with Screw Cap (850530O-200mg)
5 mL PTFE Vial with Screw Cap (850530O-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa V Misquitta et al.
Structure (London, England : 1993), 12(12), 2113-2124 (2004-12-04)
Monoacylglycerols have been used as bilayered hosts for growing crystals of membrane proteins. To date, the lipids used have had chains 16 and 18 carbon atoms long. We hypothesized that a shorter-chained lipid producing a thinner bilayer would facilitate the
Protein
Johner N, et al.
Journal of the American Chemical Society, 136, 3271-3271 (2014)
Kai Yu Chen et al.
Journal of agricultural and food chemistry, 67(29), 8191-8196 (2019-07-10)
Conversion of free fatty acids into monoacylglycerol gives rise to new structural properties, particularly amphipathic property. Therefore, monoacylglycerols are widely used in pharmaceutical and food industries and are also reported to facilitate better absorption into the human body. A functional
Kumar Nagarathinam et al.
Acta crystallographica. Section F, Structural biology communications, 73(Pt 7), 423-430 (2017-07-12)
The active efflux of antibiotics by multidrug-resistance (MDR) transporters is a major pathway of drug resistance and complicates the clinical treatment of bacterial infections. MdfA is a member of the major facilitator superfamily (MFS) from Escherichia coli and provides resistance
Dianfan Li et al.
Crystal growth & design, 11(2), 530-537 (2011-07-12)
The default lipid for the bulk of the crystallogenesis studies performed to date using the cubic mesophase method is monoolein. There is no good reason however, why this 18-carbon, cis-monounsaturated monoacylglycerol should be the preferred lipid for all target membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service