Skip to Content
Merck
All Photos(2)

Key Documents

850482C

Avanti

16:0-18:0 (11-12BR) PC

1-palmitoyl-2-(11,12-dibromo)stearoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-hexadecanoyl-2-(11,12-dibromooctadecanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$879.00

$879.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$879.00

About This Item

Empirical Formula (Hill Notation):
C42H82NO8PBr2
CAS Number:
Molecular Weight:
919.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$879.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (850482C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850482C

concentration

10 mg/mL (850482C-10mg)

shipped in

dry ice

storage temp.

−20°C

General description

16:0-18:0 (11-12BR) PC (1-palmitoyl-2-(11,12-dibromo) stearoyl-sn-glycero-3-phosphocholine) consists of brominated stearoyl chains.[1] This brominated-PC lipid[2] is non-hygroscopic and has a stability of 1 year.

Application

16:0-18:0 (11-12BR) PC (1-palmitoyl-2-(11,12-dibromo) stearoyl-sn-glycero-3-phosphocholine) may be used:
  • in phospholipid vesicle preparation for tryptophan (Trp) penetration depth experiments[2]
  • to determine the bromination effect at different positions along the fatty acid chains[3]
  • for quenching of human heat shock protein 70 (Hsp70) tryptophan (Trp) fluorescence[1]

Biochem/physiol Actions

16:0-18:0 (11-12BR) PC (1-palmitoyl-2-(11,12-dibromo) stearoyl-sn-glycero-3-phosphocholine) can be used in the preparation of vesicles.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850482C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ajay K Mahalka et al.
Biochimica et biophysica acta, 1838(5), 1344-1361 (2014-02-01)
While a significant fraction of heat shock protein 70 (Hsp70) is membrane associated in lysosomes, mitochondria, and the outer surface of cancer cells, the mechanisms of interaction have remained elusive, with no conclusive demonstration of a protein receptor. Hsp70 contains
Tim Rasmussen et al.
Journal of molecular biology, 431(17), 3339-3352 (2019-06-08)
All membrane proteins have dynamic and intimate relationships with the lipids of the bilayer that may determine their activity. Mechanosensitive channels sense tension through their interaction with the lipids of the membrane. We have proposed a mechanism for the bacterial
Candace M Pfefferkorn et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2821-2828 (2015-08-25)
Apolipoproteins are essential human proteins for lipid metabolism. Together with phospholipids, they constitute lipoproteins, nm to μm sized particles responsible for transporting cholesterol and triglycerides throughout the body. To investigate specific protein-lipid interactions, we produced and characterized three single-Trp containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service