Skip to Content
Merck
All Photos(2)

Key Documents

850481C

Avanti

16:0-18:0 (9-10BR) PC

1-palmitoyl-2-(9,10-dibromo)stearoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-hexadecanoyl-2-(9,10-dibromooctadecanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$926.00

$926.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$926.00

About This Item

Empirical Formula (Hill Notation):
C42H82NO8PBr2
CAS Number:
Molecular Weight:
919.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$926.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (850481C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850481C

concentration

10 mg/mL (850481C-10mg)

shipped in

dry ice

storage temp.

−20°C

General description

16:0-18:0 (9-10BR) PC (1-palmitoyl-2-(9,10-dibromo) stearoyl-sn-glycero-3-phosphocholine) is a brominated phosphatidylcholine lipid.[1] This lipid is stable for one year.

Application

16:0-18:0 (9-10BR) PC (1-palmitoyl-2-(9,10-dibromo) stearoyl-sn-glycero-3-phosphocholine) has been used in large unilamellar vesicles (LUVs) and small unilamellar vesicles (SUVs) for fluorescence quenching.[2] It has also been used to prepare large unilamellar vesicles (LUV) for liposome co-sedimentation assay.[3] It may be used as a component in large unilamellar liposomes for parallax analysis[4] and in lipid vesicle preparation for tryptophan (Trp) penetration depth experiments/bromine quenching experiments.[1][5]

Biochem/physiol Actions

16:0-18:0 (9-10BR) PC (1-palmitoyl-2-(9,10-dibromo) stearoyl-sn-glycero-3-phosphocholine) can act as a deep quencher.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850481C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Montagner et al.
Biochemistry, 46(7), 1878-1887 (2007-01-26)
During intoxication of a cell, the translocation (T) domain of the diphtheria toxin helps the passage of the catalytic domain across the membrane of the endosome into the cytoplasm. We have investigated the behavior of the N-terminal region of the
Thai Leong Yap et al.
The Journal of biological chemistry, 290(2), 744-754 (2014-11-28)
Mutations in glucocerebrosidase (GCase), the enzyme deficient in Gaucher disease, are a common genetic risk factor for the development of Parkinson disease and related disorders, implicating the role of this lysosomal hydrolase in the disease etiology. A specific physical interaction
Kwok-Ho Lam et al.
Cell reports, 30(8), 2526-2539 (2020-02-27)
Botulinum neurotoxin (BoNT) is one of the most acutely lethal toxins known to humans, and effective treatment for BoNT intoxication is urgently needed. Single-domain antibodies (VHH) have been examined as a countermeasure for BoNT because of their high stability and
John M Harrington et al.
PloS one, 7(9), e44384-e44384 (2012-09-13)
The bloodstream developmental forms of pathogenic African trypanosomes are uniquely susceptible to killing by small hydrophobic peptides. Trypanocidal activity is conferred by peptide hydrophobicity and charge distribution and results from increased rigidity of the plasma membrane. Structural analysis of lipid-associated
Candace M Pfefferkorn et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2821-2828 (2015-08-25)
Apolipoproteins are essential human proteins for lipid metabolism. Together with phospholipids, they constitute lipoproteins, nm to μm sized particles responsible for transporting cholesterol and triglycerides throughout the body. To investigate specific protein-lipid interactions, we produced and characterized three single-Trp containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service