Skip to Content
Merck
All Photos(2)

Key Documents

850367C

Avanti

19:0 PC

1,2-dinonadecanoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

PC(19:0/19:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$242.00

$242.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$242.00

About This Item

Empirical Formula (Hill Notation):
C46H92NO8P
CAS Number:
Molecular Weight:
818.20
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$242.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850367C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850367C

concentration

10 mg/mL (850367C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Phosphocholine is considered as a precursor molecule of phosphatidylcholine.[1] 19:0 PC is a linear saturated acyl chain with 19 carbon atoms.[2]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

19:0 PC may be used:
  • as an internal standard to extract total lipids from human brain region homogenates[3]
  • in multilamellar vesicles for biophysical studies[2]
  • as an internal standard for soil microbial community analysis[4]

Packaging

5 mL Clear Glass Sealed Ampule (850367C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey A Potekhin et al.
Biochimica et biophysica acta, 1808(7), 1806-1810 (2011-03-08)
The bilayer phase transitions from the ripple gel phase (P'(β)) to the liquid-crystal phase (L(α)) of a series of 1,2-diacylphosphatidylcholines containing a linear saturated acyl chain (C=14-19) have been studied by high-pressure scanning microcalorimetry. It has been shown that at
Alba Naudí et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1862(5), 485-495 (2017-02-12)
Lipids played a determinant role in the evolution of the brain. It is postulated that the morphological and functional diversity among neural cells of the human central nervous system (CNS) is projected and achieved through the expression of particular lipid
Polyploidy in invasive Solidago canadensis increased plant nitrogen uptake, and abundance and activity of microbes and nematodes in soil
Wu S, et al.
Soil Biology and Biochemistry, 138, 107594-107594 (2019)
Biology-cancer metabolic phenotype
NMR Metabolomics in Cancer Research, 15-138 (2013)
Hsiu-Ching Hsu et al.
Journal of biomedical science, 21, 101-101 (2014-11-20)
Use of the chemotherapeutic drug doxorubicin (DOX) is associated with serious cardiotoxicity, as it increases levels of reactive oxygen species (ROS). N-3 polyunsaturated fatty acid dietary supplements can be of benefit to patients undergoing cancer therapy. The aims of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service