Skip to Content
Merck
All Photos(2)

Key Documents

850303P

Avanti

04:0 PC

1,2-dibutyryl-sn-glycero-3-phosphocholine, powder

Synonym(s):

PC(4:0/4:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$209.00

$209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$209.00

About This Item

Empirical Formula (Hill Notation):
C16H32NO8P
CAS Number:
Molecular Weight:
397.40
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$209.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850303P-25mg)
pkg of 1 × 500 mg (850303P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850303P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCC)=O)COC(CCC)=O)=O

InChI

1S/C16H32NO8P/c1-6-8-15(18)22-12-14(25-16(19)9-7-2)13-24-26(20,21)23-11-10-17(3,4)5/h14H,6-13H2,1-5H3/t14-/m1/s1

InChI key

QIJYAMAPPUXBSC-CQSZACIVSA-N

General description

04:0 PC is a water soluble phophatidylcholine analogue.[1] Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes.[2] It is also an important component of the mucosal layer of the colon.[3]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

04:0 PC is suitable for use to evaluate its ability to form reverse micelles.[4] It is also suitable for use as a phospholipid substrate to study the substrate specificity of glycerophosphoethanolamine ethanolaminephosphodiesterase (GPE-EP).[5]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[3] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[6]

Packaging

30 mL Amber Wide Mouth Bottle with Screw Cap (850303P-500mg)
5 mL Clear Glass Sealed Ampule (850303P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of glycerophosphoethanolamine ethanolaminephosphodiesterase from Streptomyces sanglieri
Mineta S, et al.
Journal of Bioscience and Bioengineering, 119(2), 123-130 (2015)
Integrative Medicine - E-Book (2017)
Role of reverse micelles on lipid oxidation in bulk oils: impact of phospholipids on antioxidant activity of alpha-tocopherol and Trolox
Chen B, et al.
Food & Function, 2(6), 302-309 (2011)
The Membranes of Cells (2016)
F Berr et al.
The Journal of biological chemistry, 268(6), 3976-3979 (1993-02-25)
In the present study we used the water-soluble short chain phosphatidylcholine analogue L-alpha-dibutyryl-glycero-3-phosphatidylcholine (diC4PC) to investigate the mechanism involved in the canalicular secretion of phospholipids in rat liver. Uptake of 14C-labeled di-C4PC was studied in isolated microsomes as well as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service