Skip to Content
Merck
All Photos(2)

Key Documents

850116P

Avanti

IP4(1,3,4,5)

Avanti Research - A Croda Brand 850116P, powder

Synonym(s):

D-myo-inositol-1,3,4,5-tetraphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$201.00
500 μG
$895.00

$201.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$201.00
500 μG
$895.00

About This Item

Empirical Formula (Hill Notation):
C6H28N4O18P4
CAS Number:
Molecular Weight:
568.20
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$201.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850116P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850116P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850116P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]([C@H](O)[C@H](OP(O)([O-])=O)[C@@H](OP([O-])(O)=O)[C@@H]1OP([O-])(O)=O)OP(O)([O-])=O.[NH4+].[NH4+].[NH4+].[NH4+]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850116P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850116P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service