Skip to Content
Merck
All Photos(3)

Key Documents

850091P

Avanti

Liver Lyso PI

Avanti Research - A Croda Brand 850091P, powder

Synonym(s):

L-α-lysophosphatidylinositol (Liver, Bovine) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$769.00
10 MG
$1,300.00

$769.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$769.00
10 MG
$1,300.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$769.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPI; may contain up to 10% of the 2-LPI isomer, TLC)

form

powder

packaging

pkg of 1 × 10 mg (850091P-10mg)
pkg of 1 × 5 mg (850091P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850091P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C27H53O12P.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(29)37-18-20(28)19-38-40(35,36)39-27-25(33)23(31)22(30)24(32)26(27)34;/h20,22-28,30-34H,2-19H2,1H3,(H,35,36);/q;+1/p-1/t20-,22-,23-,24+,25-,26-,27-;/m1./s1

InChI key

AHPIBXQPCBITBP-FJYNCLFYSA-M

General description

Lysophosphotidylinositol (LPI) is a bioactive lipid obtained from the phospholipase A (PLA) family of lipases.[1]

Application

Liver Lyso PI has been used to enhance the binding efficiency of ovalbumin peptide to class II major histocompatibility molecules.[2]

Biochem/physiol Actions

Lysophosphotidylinositol (LPI) is involved in metabolism and glucose homeostasis.[1] It also affects cellular growth, differentiation and motility of various cells, such as nerve cells, endothelial cells and cancer cells.[1] LPI is an endogenous ligand for G-protein coupled receptor, GPR55.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (850091P-10mg)
5 mL Amber Glass Screw Cap Vial (850091P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The lysophosphatidylinositol receptor GPR55 modulates pain perception in the periaqueductal gray
Deliu E, et al.
Molecular Pharmacology, 88(2), 265-272 (2015)
Lysophosphatidylinositol signalling and metabolic diseases
Arifin S A and Falasca M
Metabolites, 6(1), 6-6 (2016)
Phospholipids enhance the binding of peptides to class II major histocompatibility molecules
Roof R W, et al.
Proceedings of the National Academy of Sciences of the USA, 87(5), 1735-1739 (1990)
Ieva Ailte et al.
Scientific reports, 6, 30336-30336 (2016-07-28)
Shiga toxin (Stx), an AB5 toxin, binds specifically to the neutral glycosphingolipid Gb3 at the cell surface before being transported into cells. We here demonstrate that addition of conical lysophospholipids (LPLs) with large head groups inhibit Stx binding to cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service