Skip to Content
Merck
All Photos(3)

Key Documents

841148C

Avanti

Soy PG

Avanti Research - A Croda Brand 841148C

Synonym(s):

L-α-phosphatidylglycerol (Soy) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$978.00
200 MG
$2,860.00

$978.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$978.00
200 MG
$2,860.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$978.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (841148C-25mg)
pkg of 2 × 4 mL (841148C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 841148C

concentration

10 mg/mL (841148C-25mg)
25 mg/mL (841148C-200mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C40H75O10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,37-38,41-42H,3-10,12,14-16,19-36H2,1-2H3,(H,45,46);/q;+1/p-1/b13-11-,18-17-;/t37?,38-;/m1./s1

InChI key

JBOGSUDWCXWRFD-YBBFZHODSA-M

General description

Soy PG is a mixture of phosphatidylglycerol (PG) with a high proportion of polyunsaturated fatty acids.[1]

Application

Soy PG has been used for lipid-protein overlay screen assay.[2]

Biochem/physiol Actions

Soy PG prevents keratinocyte proliferation and is used to treat psoriasis.[1]

Packaging

5 mL Clear Glass Sealed Ampule (841148C-200mg)
5 mL Clear Glass Sealed Ampule (841148C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soy phosphatidylglycerol reduces inflammation in a contact irritant ear edema mouse model in vivo
Xie D, et al.
Journal of Pharmacology and Experimental Therapeutics, 366(1) (2018)
Activation of ROP6 GTPase by phosphatidylglycerol in Arabidopsis
Han X, et al.
Frontiers in Plant Science, 9(1), 347-347 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service