Skip to Content
Merck
All Photos(3)

Key Documents

840044C

Avanti

Soy PI

Avanti Research - A Croda Brand 840044C

Synonym(s):

L-α-phosphatidylinositol (Soy) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$510.00
25 MG
$914.00
50 MG
$1,520.00

$510.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$510.00
25 MG
$914.00
50 MG
$1,520.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$510.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (840044C-10mg)
pkg of 1 × 2 mL (840044C-50mg)
pkg of 1 × 2.5 mL (840044C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840044C

concentration

10 mg/mL (840044C-10mg)
10 mg/mL (840044C-25mg)
25 mg/mL (840044C-50mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]([C@H](O)[C@H](O)[C@@H](O)[C@@H]1O)OP([O-])(OC[C@@]([H])(COC(CCCCCCCCCCCCCCC)=O)OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)=O.[Na+]

InChI

1S/C43H79O13P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(45)55-35(34-54-57(51,52)56-43-41(49)39(47)38(46)40(48)42(43)50)33-53-36(44)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,35,38-43,46-50H,3-10,12,14-16,19-34H2,1-2H3,(H,51,52);/q;+1/p-1/b13-11-,18-17-;/t35-,38?,39-,40?,41?,42?,43-;/m1./s1

InChI key

DCDQYHYLAGWAPO-UQZYTPKFSA-M

General description

L-α-phosphatidylinositol (Soy) is a naturally occurring, negatively charged glycerophospholipid with an inositol ring.[1] It is found in biological membranes, which are involved in scaffolding, cellular compartmentalization and signal transduction.[1]

Application

Soy PI has been used in PI 3-kinase assay.[2]

Biochem/physiol Actions

L-α-phosphatidylinositol (Soy) acts as a cellular mediator in signal transduction processes. It is used to design therapeutic delivery vehicles for drug delivery.[3] Soy PI also contributes to the regulation of cholesterol metabolism.[4]

Packaging

5 mL Clear Glass Sealed Ampule (840044C-10mg)
5 mL Clear Glass Sealed Ampule (840044C-25mg)
5 mL Clear Glass Sealed Ampule (840044C-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vps34 kinase domain dynamics regulate the autophagic PI 3-kinase complex
Stjepanovic G, et al.
Molecular Cell, 67(3), 528-534 (2017)
Phospholipids as cardiovascular therapeutics
Pandey NR and Sparks DL
Current Opinion in Investigational Drugs, 9(3), 281-285 (2008)
Aaron Peng et al.
Chemistry and physics of lipids, 165(1), 15-22 (2011-10-26)
Liposomes consisted of phosphatidylinositol (PI) and phosphatidylcholine (PC) have been utilized as delivery vehicle for drugs and proteins. In the present work, we studied the effect of soy PI on physical properties of 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) liposomes such as phase state
Reorganization of Ternary Lipid Mixtures of Nonphosphorylated Phosphatidylinositol Interacting with Angiomotin
Kimble-Hill, et al.
The Journal of Physical Chemistry B, 122(35), 8404-8415 (2018)
Ashutosh Tripathi et al.
The Journal of biological chemistry, 294(50), 19081-19098 (2019-11-07)
Phosphatidylinositol-transfer proteins (PITPs) are key regulators of lipid signaling in eukaryotic cells. These proteins both potentiate the activities of phosphatidylinositol (PtdIns) 4-OH kinases and help channel production of specific pools of phosphatidylinositol 4-phosphate (PtdIns(4)P) dedicated to specific biological outcomes. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service