Skip to Content
Merck
All Photos(3)

Key Documents

840012C

Avanti

Heart CA

Avanti Research - A Croda Brand

Synonym(s):

Cardiolipin (Heart, Bovine) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$278.00
50 MG
$458.00
100 MG
$769.00

$278.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$278.00
50 MG
$458.00
100 MG
$769.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$278.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2 mL (840012C-50mg)
pkg of 1 × 2.5 mL (840012C-25mg)
pkg of 1 × 4 mL (840012C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840012C-25mg)
25 mg/mL (840012C-100mg)
25 mg/mL (840012C-50mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCC\C=C/C\C=C/CCCCC)OCC(O)CO[P](=O)([O-])OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCC\C=C/C\C=C/CCCCC

InChI

1S/C81H142O17P2.2Na/c1-5-9-13-17-21-25-29-33-37-41-45-49-53-57-61-65-78(83)91-71-76(97-80(85)67-63-59-55-51-47-43-39-35-31-27-23-19-15-11-7-3)73-95-99(87,88)93-69-75(82)70-94-100(89,90)96-74-77(98-81(86)68-64-60-56-52-48-44-40-36-32-28-24-20-16-12-8-4)72-92-79(84)66-62-58-54-50-46-42-38-34-30-26-22-18-14-10-6-2;;/h21-28,33-40,75-77,82H,5-20,29-32,41-74H2,1-4H3,(H,87,88)(H,89,90);;/q;2*+1/p-2/b25-21-,26-22-,27-23-,28-24-,37-33-,38-34-,39-35-,40-36-;;/t76-,77-;;/m1../s1

InChI key

ZGSPNIOCEDOHGS-LBOJVHLDSA-L

General description

Cardiolipin is a phospholipid found in the inner membrane of the mitochondria. It is synthesized from phosphatidylglycerol and cytidinediphosphate-diacylglycerol.[1] It consists of two phosphatidyl residues, which are linked by a glycerol group.[2]

Application

Heart CA has been used in the Mycobacterium lipid extraction and in β- Hexosaminidase release and calcium mobilization assays.[3] It maybe used for liposome synthesis[4]. It may also be used as a lipid standard for the in situ analysis and characterization of cardiolipins.[5]

Biochem/physiol Actions

Cardiolipin is involved in maintaining the structure and function of the mitochondria. It also serves as a co-factor for stimulating mitochondrial enzymes and mediates mitochondrial apoptosis. Cardiolipin is associated with the oxidative phosphorylation system for the adenine triphosphate (ATP) synthesis in the inner membrane of the mitochondria.[1] Deficiency of cardiolipin is associated with variation of mitochondrial membrane structure and function. It is also associated with Barth syndrome.[2][1]

Packaging

5 mL Clear Glass Sealed Ampule (840012C-100mg)
5 mL Clear Glass Sealed Ampule (840012C-25mg)
5 mL Clear Glass Sealed Ampule (840012C-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cardiolipin, the heart of mitochondrial metabolism
Houtkooper R H and Vaz F M
Cellular and Molecular Life Sciences, 65(16), 2493-2506 (2008)
Direct MALDI-MS analysis of cardiolipin from rat organs sections
Wang Hay-Yan J, et al.
Journal of the American Society For Mass Spectrometry, 18(3), 567-577 (2007)
Cardiolipin stabilizes respiratory chain supercomplexes
Pfeiffer K, et al.
The Journal of Biological Chemistry, 278(52), 52873-52880 (2003)
Mycobacterial Lipids Induce Calcium Mobilization and Degranulation of Mast Cells
Torres-Atencio I, et al.
American Journal of Respiratory and Critical Care Medicine, 198(6), 813-816 (2018)
Mitochondrial translocation of alpha-synuclein is promoted by intracellular acidification
Cole NB, et al.
Experimental Cell Research, 314(10), 2076-2089 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service