Skip to Content
Merck
All Photos(1)

Key Documents

810892C

Avanti

DSPE PEG(2000)-N-Cyanine 7

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[amino(polyethylene glycol)-2000]-N-(Cyanine 7)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$912.00

$912.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$912.00

About This Item

Empirical Formula (Hill Notation):
C169H305N4O55P
CAS Number:
Molecular Weight:
3304.20
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$912.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mg (
810892C-1MG)

pkg of 5 × 1 mg (
810892C-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (810892C-1MG)
1 mg/mL (810892C-5MG)

shipped in

dry ice

storage temp.

−20°C

General description

1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine-polyethylene glycol 2000 (DSPE PEG 2000) are oblate spheroids in aqueous solution. The diameter ranges from 18nm to 90nm and are effective agents for drug delivery.[1]

Packaging

5ML Clear Glass Sealed Ampule (810892C-1MG)
5ML Clear Glass Sealed Ampule (810892C-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of the lipid chain melting transition on the stability of DSPE-PEG (2000) micelles
Kastantin M, et al.
Langmuir, 25(13), 7279-7286 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service