Skip to Content
Merck
All Photos(1)

Documents

810386C

Avanti

18:1 PE-TopFluor AF488

Avanti Research - A Croda Brand

Synonym(s):

1,2-DIOLEOYL-SN-GLYCERO-3-PHOSPHOETHANOLAMINE-N-( TopFluor AF488) (AMMONIUM SALT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H99N6O18PS2
CAS Number:
Molecular Weight:
1311.58
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 250 μg (810386C-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

250 μg/mL (810386C-250ug)

shipped in

dry ice

storage temp.

−20°C

Packaging

5ml Amber Glass Screw Cap Vial (810386C-250UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service