Skip to Content
Merck
All Photos(1)

Key Documents

810346C

Avanti

18:0 Cyanine 5.5 PE

Avanti Research - A Croda Brand 810346C

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-(Cyanine 5.5)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$895.00

$895.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$895.00

About This Item

Empirical Formula (Hill Notation):
C81H126ClN4O9P
CAS Number:
Molecular Weight:
1366.32
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$895.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810346C-1mg)
pkg of 5 × 1 mL (810346C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810346C

concentration

1 mg/mL (810346C-1mg)
1 mg/mL (810346C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:0 Cy5.5 PE or 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-(Cyanine 5.5) is a phosphoethanolamine (PE) derivative wherein the amine and hydroxyl groups of PE are substituted with cyanine 5.5 and distearoyl-sn-glycerol.

Application

18:0 Cy5.5 PE has been used in the preparation of fluorescently labelled liposomes, for direct visualization of nitrobenzoxadiazole (NBD)-palmitoyl-CoA association with a liposome.[1] It has also been used to fluorescently label small unilamellar vesicles (SUV) and large unilamellar vesicles (LUV) to study the interaction of viral particles with phospholipids.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810346C-1mg)
5 mL Amber Glass Screw Cap Vial (810346C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Oleksandr Kolyvushko et al.
Pathogens (Basel, Switzerland), 9(5) (2020-05-14)
Exposure of phosphatidylserine (PS) in the outer leaflet of the plasma membrane is induced by infection with several members of the Alphaherpesvirinae subfamily. There is evidence that PS is used by the equine herpesvirus type 1 (EHV-1) during entry, but
James J Asciolla et al.
Cell reports, 29(13), 4608-4619 (2019-12-26)
Attachment of palmitate to the N terminus of Sonic hedgehog (Shh) is essential for Shh signaling. Shh palmitoylation is catalyzed on the luminal side of the endoplasmic reticulum (ER) by Hedgehog acyltransferase (Hhat), an ER-resident enzyme. Palmitoyl-coenzyme A (CoA), the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service