Skip to Content
Merck
All Photos(1)

Key Documents

810288P

Avanti

16:0 TopFluor cholesterol

Avanti Research - A Croda Brand 810288P, powder

Synonym(s):

23-(dipyrrometheneboron difluoride)-24-norcholesteryl palmitate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$655.00

$655.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$655.00

About This Item

Empirical Formula (Hill Notation):
C52H81 BF2N2O2
CAS Number:
Molecular Weight:
815.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$655.00


Estimated to ship on16 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810288P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810288P

shipped in

dry ice

storage temp.

−20°C

General description

16:0 TopFluor cholesterol carries saturated acyl chains. Approximately 30-40% of the mammalian plasma membrane consists of cholesterol.[1] TopFluor has a moiety, called dipyrromethene boron difluoride. This fluorescent molecule is covalently attached to various lipid molecules.[2]

Application

16:0 TopFluor cholesterol may be used to study its diffusion with phospholipid analogs in model membrane in order to understand cholesterol dynamics at the nanoscale in biomembranes.[1]

Biochem/physiol Actions

Cholesterol participates in various signaling processes. It also helps in shaping the structure of molecular membrane.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810288P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoscale dynamics of cholesterol in the cell membrane
Pinkwart K, et al.
The Journal of biological chemistry, 294(34), 12599-12609 (2019)
Investigation of the biophysical properties of a fluorescently modified ceramide-1-phosphate
Shirey CM, et al.
Chemistry and Physics of Lipids, 200, 32-32 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service