Skip to Content
Merck
All Photos(1)

Key Documents

810203P

Avanti

NBD Lyso SM

Avanti Research - A Croda Brand 810203P, powder

Synonym(s):

omega(7-nitro-2-1,3-benzoxadiazol-4-yl)-D-erythro-sphingosine-1-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C293H51N6O8P
CAS Number:
Molecular Weight:
3813.55
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (with stopper and crimp cap (810203P-50ug))

manufacturer/tradename

Avanti Research - A Croda Brand 810203P

shipped in

dry ice

storage temp.

−20°C

General description

Lyso-sphingomyelin is a lysospingolipid used as a diagnostic biomarker. It allows a clear distinction between Niemann-Pick C disease (NP-C) and acid sphingomyelinase deficiency (ASMD).[1]

Application

NBD Lyso SM or omega(7-nitro-2-1,3-benzoxadiazol-4-yl)-D-erythro-sphingosine-1-phosphocholine is suitable to determine the hydrolysis of sphingomyelin (SM) by RSp1609: acid sphingomyelinase-like phosphodiesterase (RsASML) protein.[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810203P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The impact of biomarkers analysis in the diagnosis of Niemann-Pick C disease and acid sphingomyelinase deficiency
Deodato F, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 486, 387-394 (2018)
Federica Deodato et al.
Clinica chimica acta; international journal of clinical chemistry, 486, 387-394 (2018-08-29)
Although representing two distinct disease entities, Niemann-Pick disease type C (NP-C) disease and acid sphingomyelinase deficiency (ASMD) share several phenotypic features. The lack of biomarkers was responsible in the past of diagnostic delay. Recently, plasma oxysterols, cholestan-3β,5α,6β-triol (Triol) and 7-ketocholesterol
Michael V Airola et al.
PloS one, 9(8), e105830-e105830 (2014-08-22)
Acid sphingomyelinase (aSMase) is a human enzyme that catalyzes the hydrolysis of sphingomyelin to generate the bioactive lipid ceramide and phosphocholine. ASMase deficiency is the underlying cause of the genetic diseases Niemann-Pick Type A and B and has been implicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service