Skip to Content
Merck
All Photos(1)

Key Documents

810197P

Avanti

16:0-Pyr PC

Avanti Research - A Croda Brand 810197P, powder

Synonym(s):

1-hexadecanoyl-2-(1-pyrenedecanoyl)-sn-glycero-3-phosphatidylcholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$352.00

$352.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$352.00

About This Item

Empirical Formula (Hill Notation):
C50H76NO8P
CAS Number:
Molecular Weight:
850.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$352.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810197P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810197P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCc1c2c3c4c(cc2)cccc4ccc3cc1)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C50H76NO8P/c1-5-6-7-8-9-10-11-12-13-14-17-20-23-29-47(52)56-39-45(40-58-60(54,55)57-38-37-51(2,3)4)59-48(53)30-24-21-18-15-16-19-22-26-41-31-32-44-34-33-42-27-25-28-43-35-36-46(41)50(44)49(42)43/h25,27-28,31-36,45H,5-24,26,29-30,37-40H2,1-4H3/t45-/m1/s1

InChI key

ZZLOWUUVDRBGKM-WBVITSLISA-N

General description

Phosphatidylcholine (PC) is a common phospholipid found in eukaryotes and a prime metabolite of glycerolipid metabolism.[1] It is a vital component of the mucosal layer of the colon.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810197P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Importance of phosphatidylcholine on the chloroplast surface
Botella C, et al.
Chemistry and Physics of Lipids null
The critical role of phosphatidylcholine and phosphatidylethanolamine metabolism in health and disease
van der Veen JN, et al.
Biochimica et biophysica acta. General subjects null
César Botella et al.
Progress in lipid research, 65, 12-23 (2016-11-23)
In plant cells, phosphatidylcholine (PC) is a major glycerolipid of most membranes but practically lacking from the plastid internal membranes. In chloroplasts, PC is absent from the thylakoids and the inner envelope membrane. It is however the main component of
Jelske N van der Veen et al.
Biochimica et biophysica acta. Biomembranes, 1859(9 Pt B), 1558-1572 (2017-04-16)
Phosphatidylcholine (PC) and phosphatidylethanolamine (PE) are the most abundant phospholipids in all mammalian cell membranes. In the 1950s, Eugene Kennedy and co-workers performed groundbreaking research that established the general outline of many of the pathways of phospholipid biosynthesis. In recent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service