Skip to Content
Merck
All Photos(2)

Key Documents

810154P

Avanti

16:0-12:0 NBD PE

Avanti Research - A Croda Brand 810154P, powder

Synonym(s):

1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$484.00

$484.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$484.00

About This Item

Empirical Formula (Hill Notation):
C39H68N5O11P
CAS Number:
Molecular Weight:
813.96
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$484.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810154P-1mg)
pkg of 5 × 1 mg (810154P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810154P

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is a fluorescent probe localized at the membrane interface.[1][2] It possess fluorescent NBD label attached to the headgroup of phosphatidylethanolamine (PE).[2] PE is the parent compound for phosphatidylcholine and phosphatidylethanolamine synthesis[3] and is mostly enriched in brain.[4]

Application

16:0-12:0 NBD PE has been used in lipid extraction assay and in vitro phospholipid fluorescence resonance energy transfer (FRET) assays.[5] It is suitable for the determination of enzyme substrate specificity of lipoteichoic acid synthase (LtaS).[6][7][6]

Biochem/physiol Actions

Phosphoethanolamine (PE) plays a vital role in the prevention of tumor proliferation.[3] N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) monitors the Red edge excitation shift (REES) phenomenon with the unique motional and dielectric properties it possesses.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810154P-1mg)
5 mL Amber Glass Screw Cap Vial (810154P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theoretical Study of Phosphoethanolamine: A Synthetic Anticancer Agent with Broad Antitumor Activity
Lorenzo VP, et al.
Journal of Chemistry, 2016 (2016)
Application of NBD-labeled lipids in membrane and cell biology
Fluorescent methods to study biological membranes, 37-50 (2012)
Ouma Onguka et al.
The Journal of biological chemistry, 290(20), 12744-12752 (2015-04-02)
Phosphatidylethanolamine (PE) is a major cellular phospholipid that can be made by four separate pathways, one of which resides in the mitochondrion. The mitochondrial enzyme that generates PE is phosphatidylserine decarboxylase 1 (Psd1p). The pool of PE produced by Psd1p
Jean E Vance et al.
Biochimica et biophysica acta, 1831(3), 543-554 (2012-09-11)
Phosphatidylserine (PS) and phosphatidylethanolamine (PE) are metabolically related membrane aminophospholipids. In mammalian cells, PS is required for targeting and function of several intracellular signaling proteins. Moreover, PS is asymmetrically distributed in the plasma membrane. Although PS is highly enriched in
Maria Mercedes Palomino et al.
Microbiology (Reading, England), 159(Pt 11), 2416-2426 (2013-09-10)
The probiotic Gram-positive bacterium Lactobacillus casei BL23 is naturally confronted with salt-stress habitats. It has been previously reported that growth in high-salt medium, containing 0.8 M NaCl, leads to modifications in the cell envelope of this bacterium. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service