Skip to Content
Merck
All Photos(1)

Key Documents

800817C

Avanti

18:0-18:2 DG

1-stearoyl-2-linoleoyl-sn-glycerol, chloroform

Synonym(s):

1-octadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycerol; DG(18:0/18:2(9Z,12Z)/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$875.00

$875.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$875.00

About This Item

Empirical Formula (Hill Notation):
C39H72O5
CAS Number:
Molecular Weight:
620.99
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$875.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (800817C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800817C

concentration

2 mg/mL (800817C-10mg)

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O([C@H](COC(=O)CCCCCCCCCCCCCCCCC)CO)C(=O)CCCCCCC\C=C/C\C=C/CCCCC

InChI

1S/C39H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h12,14,18,20,37,40H,3-11,13,15-17,19,21-36H2,1-2H3/b14-12-,20-18-/t37-/m0/s1

InChI key

AJMZUFBKADIAKC-SKTOPKGQSA-N

General description

18:0-18:2 DG (diacylglycerol) is a second messenger signaling lipid in biochemical signaling and is a product of the hydrolysis of the phospholipid PIP2 (phosphatidylinositolbisphosphate) by the enzyme phospholipase C (PLC) (a membrane-bound enzyme) that, through the same reaction, produces inositol trisphosphate (IP3). It possess hydrophobic properties due to which it remains within the plasma membrane.

Biochem/physiol Actions

DAG plays a vital role in the physiological activation of protein kinase C (PKC). The production of DAG in the membrane facilitates translocation of PKC from the cytosol to the plasma membrane.[1][2][3][4] Diacylglycerol mimicks the effects of the tumor-promoting compounds phorbol esters.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800817C-10mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells:
Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration:
Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Specificity and Mechanism of Protein Kinase C Activation by sn-1,2-diacylglycerols.
Ganong BR, et al.
Proceedings of the National Academy of Sciences of the USA, 83, 1184-1188 (1986)
J G Ebeling et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(3), 815-819 (1985-02-01)
Activation of cellular protein kinase C appears to be involved in the mechanism by which phorbol diesters induce differentiation of human myeloid leukemia cells (HL-60). Protein kinase C is thought to be physiologically activated by diacylglycerol derived from receptor-mediated phosphatidylinositol
R J Davis et al.
The Journal of biological chemistry, 260(3), 1562-1566 (1985-02-10)
The cell-permeable diacylglycerol, sn-1,2-dioctanoylglycerol (DiC8), is shown to mimic the effect of tumor promoting phorbol diesters on epidermal growth factor (EGF) binding and action in intact cells. DiC8 inhibited the binding of [3H]phorbol dibutyrate to A431 cell monolayers indicating that
E G Lapetina et al.
The Journal of biological chemistry, 260(3), 1358-1361 (1985-02-10)
The ability of exogenous sn-1,2-diacylglycerols and analogs to function as bioregulators of protein kinase C in human platelets was investigated. The activation of protein kinase C in platelets is indicated by specific phosphorylation of a 40,000-dalton protein. Dihexanoylglycerol, dioctanoylglycerol (diC8)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service