Skip to Content
Merck
All Photos(1)

Key Documents

800715P

Avanti

N-P Serine PA

Avanti Research - A Croda Brand 800715P, powder

Synonym(s):

N-palmitoyl-serine phosphoric acid (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$304.00

$304.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$304.00

About This Item

Empirical Formula (Hill Notation):
C19H44N3O7P
CAS Number:
Molecular Weight:
457.54
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$304.00


Estimated to ship on27 May 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (800715P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800715P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

General description

Lysophosphatidic acid (LPA) receptor modulators include N-palmitoyl serine phosphoric acid and N-palmitoyl-tyrosine phosphoric acid. N-palmitoyl serine phosphoric acid is a competitive inhibitor of the LPA receptor in Xenopus oocytes.[1][2] However, in mammalian cells, N-palmitoyl-serine phosphoric acid may act as an agonist for the LPA receptor. [3][4] LPA is a lipid mediator that acts similar to growth factors through G-protein coupled plasma membrane receptors.[1][2][3][4][5] LPA may play a role in platelet aggregation, smooth muscle contraction, vasoactive changes, cytoskeletal reorganization and cell proliferation.[1] N-palmitoyl serine phosphoric acid may also play a role in signal transduction in mammalian cells by increasing intracellular calcium. [4][5]
N-palmitoyl serine phosphoric acid prevents human platelet aggregation, which is induced by 1-hexadecyl-2-lyso-sn-glycero-3-phosphate.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (800715P-1mg)

Preparation Note

Product use: A range of 0.1-20 mM of N-palmitoyl serine phosphoric acid can be used for cell studies. Make a stock solution of 5 mM in DMSO and store at -20°C. Diluted N-palmitoyl serine phosphoric acid can be directly added to the study medium. [5]An et al. dissolved N-palmitoyl-serine and N-palmitoyl-tyrosine phosphoric acid in 0.1 mL PBS containing 0.1 mg/mL human serum albumin before adding to cells.[4] In X. laevis studies, these LPA inhibitors were dissolved in DMSO at 1 mM and filtered through a 0.45 mM membrane filter before injection.[2]

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of a receptor subtype-selective lysophosphatidic acid mimetic
Hooks, S.B, et. al.
Molecular Pharmacology, 53, 188-194 (1998)
Novel effect of N-palmitoyl-L-serine phosphoric acid on cytosolic Ca2+ levels in human osteoblasts
Jan, C.R
Pharmacology & Toxicology, 93, 71-76 (2003)
Chung-Ren Jan et al.
Pharmacology & toxicology, 93(2), 71-76 (2003-08-06)
The effect of N-palmitoyl-L-serine phosphoric acid (L-NASPA), which has been used as an inhibitor of lysophosphatidic acid receptors, on intracellular Ca2+ concentration ([Ca2+]i) in human osteosarcoma MG63 cells was measured by using fura-2. L-NASPA (0.1-10 microM) caused a rapid and
Recombinant human G protein-coupled lysophosphatidic acid receptors mediate intracellular calcium mobilization.
An, S
Molecular Pharmacology, 54, 881-888 (1998)
Inhibitors of lipid phosphatidate receptors: N-palmitoyl-serine and N-palmitoyl-tyrosine phosphoric acids.
Bittman R, et al.
Journal of Lipid Research, 37(2), 391-398 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service