Skip to Content
Merck
All Photos(1)

Key Documents

791645C

Avanti

18:1-d7-cholesterol

Avanti Research - A Croda Brand 791645C

Synonym(s):

cholest-5-en-3β-yl oleate-d7

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$458.00

$458.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
$458.00

About This Item

Empirical Formula (Hill Notation):
C45H71 D7O2
CAS Number:
Molecular Weight:
658.14
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.25

$458.00


Available to ship on22 April 2025Details


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791645C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791645C

concentration

1 mg/mL (791645C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Cholesteryl oleate is a cholesteryl ester.[1] 18:1-d7-cholesterol is a deuterated oleic acid cholesteryl ester.
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

18:1-d7-cholesterol may be used in the reconstitution of acetylated low-density lipoprotein (acLDL) liposome for trafficking studies in U2OS-SRA cells.[2]

Biochem/physiol Actions

Elevation in cholesterol ester levels and its accumulation is implicated in prostate cancer (PCa). Cholesteryl oleate, may serve as a potential biomarker for diagnosis of PCa.[3]

Packaging

5 mL Clear Glass Sealed Ampule (791645C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heart Proteomics, 967-1009 (2001)
Jia Li et al.
Scientific reports, 6, 20984-20984 (2016-02-13)
In-depth delineation of lipid metabolism in prostate cancer (PCa) is significant to open new insights into prostate tumorigenesis and progression, and provide potential biomarkers with greater accuracy for improved diagnosis. Here, we performed lipidomics and transcriptomics in paired prostate cancer
McKenna Feltes et al.
Journal of lipid research, 60(3), 707-716 (2019-01-09)
Cholesterol is an essential structural component of cellular membranes and precursor molecule for oxysterol, bile acid, and hormone synthesis. The study of intracellular cholesterol trafficking pathways has been limited in part due to a lack of suitable cholesterol analogues. Herein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service